Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 17932115-17932352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNS3
Entrez Gene ID
3790
Gene Name
potassium voltage-gated channel modifier subfamily S member 3
Gene Aliases
KV9.3
UniGene
Hs.414489
Gene Chromosome Location
Chr.2: 17877847-17932961 on Build GRCh38
Transcripts
NM_001282428.1 NM_002252.4 XM_011532825.1 XM_017004059.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3364418 c.1132C>T p.P378S Chr.2: 17932140-17932140 on Build GRCh38
7084800 c.1229C>T p.S410F Chr.2: 17932237-17932237 on Build GRCh38
7140479 c.1173C>G p.I391M Chr.2: 17932181-17932181 on Build GRCh38
350520 c.1323G>T p.R441S Chr.2: 17932331-17932331 on Build GRCh38
71272 c.1133C>T p.P378L Chr.2: 17932141-17932141 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765633261 Chr.2: 17932297-17932297 on Build GRCh38
rs3747516 Chr.2: 17932241-17932241 on Build GRCh38
rs151225864 Chr.2: 17932136-17932136 on Build GRCh38
rs749834295 Chr.2: 17932244-17932245 on Build GRCh38
rs541299086 Chr.2: 17932135-17932135 on Build GRCh38
rs750849173 Chr.2: 17932307-17932307 on Build GRCh38
rs747894491 Chr.2: 17932226-17932226 on Build GRCh38
rs770872910 Chr.2: 17932271-17932271 on Build GRCh38
rs761956499 Chr.2: 17932294-17932294 on Build GRCh38
rs775235365 Chr.2: 17932287-17932287 on Build GRCh38
rs749179312 Chr.2: 17932243-17932243 on Build GRCh38
rs1007548285 Chr.2: 17932251-17932251 on Build GRCh38
rs144718402 Chr.2: 17932156-17932156 on Build GRCh38
rs754569191 Chr.2: 17932223-17932223 on Build GRCh38
rs1036787836 Chr.2: 17932175-17932175 on Build GRCh38
rs750904358 Chr.2: 17932198-17932198 on Build GRCh38
rs775409360 Chr.2: 17932280-17932280 on Build GRCh38
rs531245454 Chr.2: 17932282-17932282 on Build GRCh38
rs998070203 Chr.2: 17932207-17932207 on Build GRCh38
rs754119495 Chr.2: 17932181-17932181 on Build GRCh38
rs1055920710 Chr.2: 17932150-17932150 on Build GRCh38
rs937664203 Chr.2: 17932141-17932141 on Build GRCh38
rs767045394 Chr.2: 17932320-17932320 on Build GRCh38
rs780940477 Chr.2: 17932224-17932224 on Build GRCh38
rs998808057 Chr.2: 17932178-17932178 on Build GRCh38
rs773202985 Chr.2: 17932154-17932154 on Build GRCh38
rs34658212 Chr.2: 17932153-17932153 on Build GRCh38
rs758979123 Chr.2: 17932199-17932199 on Build GRCh38
rs202189909 Chr.2: 17932166-17932166 on Build GRCh38
rs757706180 Chr.2: 17932186-17932186 on Build GRCh38
rs768387082 Chr.2: 17932142-17932142 on Build GRCh38
rs965219845 Chr.2: 17932189-17932189 on Build GRCh38
rs764426221 Chr.2: 17932177-17932177 on Build GRCh38
rs545834036 Chr.2: 17932159-17932159 on Build GRCh38
rs775574102 Chr.2: 17932167-17932167 on Build GRCh38
rs148982914 Chr.2: 17932172-17932172 on Build GRCh38
rs769518784 Chr.2: 17932236-17932236 on Build GRCh38
rs776245657 Chr.2: 17932151-17932151 on Build GRCh38
rs763506076 Chr.2: 17932311-17932311 on Build GRCh38
rs952440137 Chr.2: 17932229-17932229 on Build GRCh38
rs746555232 Chr.2: 17932218-17932218 on Build GRCh38
rs765827628 Chr.2: 17932197-17932197 on Build GRCh38
rs1017594800 Chr.2: 17932285-17932285 on Build GRCh38

View Full Product Details