Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184059267-184059510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTR3C
Entrez Gene ID
170572
Gene Name
5-hydroxytryptamine receptor 3C
Gene Aliases
-
UniGene
Hs.632579
Gene Chromosome Location
Chr.3: 184053047-184060673 on Build GRCh38
Transcripts
NM_130770.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1421260 c.733C>T p.R245C Chr.3: 184059448-184059448 on Build GRCh38
4572545 c.779T>C p.F260S Chr.3: 184059494-184059494 on Build GRCh38
1041902 c.734G>A p.R245H Chr.3: 184059449-184059449 on Build GRCh38
1041901 c.729C>T p.I243I Chr.3: 184059444-184059444 on Build GRCh38
6468709 c.741A>C p.P247P Chr.3: 184059456-184059456 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs569567438 Chr.3: 184059435-184059435 on Build GRCh38
rs761008474 Chr.3: 184059466-184059466 on Build GRCh38
rs529622627 Chr.3: 184059405-184059405 on Build GRCh38
rs149942453 Chr.3: 184059483-184059483 on Build GRCh38
rs764628770 Chr.3: 184059467-184059467 on Build GRCh38
rs377023957 Chr.3: 184059449-184059449 on Build GRCh38
rs546295414 Chr.3: 184059352-184059352 on Build GRCh38
rs56244916 Chr.3: 184059422-184059422 on Build GRCh38
rs528175620 Chr.3: 184059319-184059319 on Build GRCh38
rs568012082 Chr.3: 184059395-184059395 on Build GRCh38
rs879677025 Chr.3: 184059407-184059407 on Build GRCh38
rs769410829 Chr.3: 184059475-184059475 on Build GRCh38
rs746862647 Chr.3: 184059414-184059414 on Build GRCh38
rs775913590 Chr.3: 184059464-184059464 on Build GRCh38
rs890994367 Chr.3: 184059293-184059293 on Build GRCh38
rs188090891 Chr.3: 184059448-184059448 on Build GRCh38
rs749365313 Chr.3: 184059424-184059424 on Build GRCh38
rs374654829 Chr.3: 184059401-184059401 on Build GRCh38
rs776475619 Chr.3: 184059423-184059423 on Build GRCh38
rs753381285 Chr.3: 184059385-184059385 on Build GRCh38
rs745537636 Chr.3: 184059399-184059399 on Build GRCh38
rs564054724 Chr.3: 184059307-184059307 on Build GRCh38
rs754335349 Chr.3: 184059397-184059397 on Build GRCh38
rs756863604 Chr.3: 184059393-184059393 on Build GRCh38
rs901016309 Chr.3: 184059428-184059428 on Build GRCh38
rs900561037 Chr.3: 184059381-184059381 on Build GRCh38
rs771021915 Chr.3: 184059432-184059432 on Build GRCh38
rs942524503 Chr.3: 184059320-184059320 on Build GRCh38
rs141515042 Chr.3: 184059486-184059486 on Build GRCh38
rs879019544 Chr.3: 184059370-184059370 on Build GRCh38
rs1022226219 Chr.3: 184059476-184059476 on Build GRCh38
rs1039120130 Chr.3: 184059337-184059337 on Build GRCh38

View Full Product Details