Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75729374-75729599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGB4
Entrez Gene ID
3691
Gene Name
integrin subunit beta 4
Gene Aliases
CD104, GP150
UniGene
Hs.632226
Gene Chromosome Location
Chr.17: 75721328-75757819 on Build GRCh38
Transcripts
NM_000213.4 NM_001005619.1 NM_001005731.2 NM_001321123.1 XM_005257309.2 XM_005257311.4 XM_006721866.3 XM_006721867.3 XM_006721868.3 XM_006721870.3 XM_011524751.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6839537 c.734G>A p.C245Y Chr.17: 75729432-75729432 on Build GRCh38
7154339 c.703G>T p.G235C Chr.17: 75729401-75729401 on Build GRCh38
4999752 c.728C>A p.A243D Chr.17: 75729426-75729426 on Build GRCh38
6839538 c.734G>A p.C245Y Chr.17: 75729432-75729432 on Build GRCh38
1285613 c.692C>T p.A231V Chr.17: 75729390-75729390 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925658786 Chr.17: 75729577-75729577 on Build GRCh38
rs774681717 Chr.17: 75729525-75729525 on Build GRCh38
rs146923910 Chr.17: 75729406-75729406 on Build GRCh38
rs756593259 Chr.17: 75729428-75729428 on Build GRCh38
rs778293141 Chr.17: 75729432-75729432 on Build GRCh38
rs61751843 Chr.17: 75729400-75729400 on Build GRCh38
rs369922465 Chr.17: 75729464-75729464 on Build GRCh38
rs967160631 Chr.17: 75729563-75729563 on Build GRCh38
rs763724502 Chr.17: 75729471-75729471 on Build GRCh38
rs756915582 Chr.17: 75729430-75729430 on Build GRCh38
rs374647847 Chr.17: 75729439-75729439 on Build GRCh38
rs749834111 Chr.17: 75729436-75729436 on Build GRCh38
rs775115165 Chr.17: 75729465-75729465 on Build GRCh38
rs555916977 Chr.17: 75729534-75729534 on Build GRCh38
rs111888494 Chr.17: 75729437-75729437 on Build GRCh38
rs1038415538 Chr.17: 75729399-75729399 on Build GRCh38
rs781241859 Chr.17: 75729423-75729423 on Build GRCh38
rs745502016 Chr.17: 75729448-75729448 on Build GRCh38
rs373192148 Chr.17: 75729445-75729445 on Build GRCh38
rs774782708 Chr.17: 75729447-75729447 on Build GRCh38
rs1005925527 Chr.17: 75729507-75729507 on Build GRCh38
rs1024074508 Chr.17: 75729562-75729562 on Build GRCh38
rs376587489 Chr.17: 75729573-75729573 on Build GRCh38
rs941460625 Chr.17: 75729394-75729394 on Build GRCh38
rs1024140563 Chr.17: 75729402-75729402 on Build GRCh38
rs922755632 Chr.17: 75729504-75729505 on Build GRCh38
rs776379718 Chr.17: 75729478-75729478 on Build GRCh38
rs536230542 Chr.17: 75729510-75729510 on Build GRCh38
rs957039802 Chr.17: 75729446-75729446 on Build GRCh38
rs975646773 Chr.17: 75729503-75729503 on Build GRCh38
rs760299085 Chr.17: 75729470-75729470 on Build GRCh38
rs751671168 Chr.17: 75729401-75729401 on Build GRCh38
rs771906597 Chr.17: 75729453-75729453 on Build GRCh38
rs761827608 Chr.17: 75729485-75729485 on Build GRCh38
rs375136714 Chr.17: 75729425-75729425 on Build GRCh38
rs576649949 Chr.17: 75729538-75729538 on Build GRCh38

View Full Product Details