Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 202123016-202123283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR37L1
Entrez Gene ID
9283
Gene Name
G protein-coupled receptor 37 like 1
Gene Aliases
ET(B)R-LP-2, ETBR-LP-2
UniGene
Hs.132049
Gene Chromosome Location
Chr.1: 202118669-202133588 on Build GRCh38
Transcripts
NM_004767.3 XM_011510158.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5926259 c.110C>T p.T37I Chr.1: 202123073-202123073 on Build GRCh38
1127189 c.203C>T p.P68L Chr.1: 202123166-202123166 on Build GRCh38
678146 c.199T>A p.Y67N Chr.1: 202123162-202123162 on Build GRCh38
3750765 c.241C>G p.P81A Chr.1: 202123204-202123204 on Build GRCh38
6674163 c.309G>C p.R103S Chr.1: 202123272-202123272 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776016622 Chr.1: 202123121-202123121 on Build GRCh38
rs750514265 Chr.1: 202123159-202123159 on Build GRCh38
rs774875744 Chr.1: 202123230-202123230 on Build GRCh38
rs143016403 Chr.1: 202123067-202123067 on Build GRCh38
rs759064184 Chr.1: 202123123-202123123 on Build GRCh38
rs529894072 Chr.1: 202123044-202123044 on Build GRCh38
rs17854616 Chr.1: 202123235-202123235 on Build GRCh38
rs144676177 Chr.1: 202123231-202123231 on Build GRCh38
rs200257132 Chr.1: 202123172-202123172 on Build GRCh38
rs892029362 Chr.1: 202123137-202123137 on Build GRCh38
rs201065708 Chr.1: 202123165-202123165 on Build GRCh38
rs145080702 Chr.1: 202123036-202123036 on Build GRCh38
rs758268638 Chr.1: 202123174-202123174 on Build GRCh38
rs771774196 Chr.1: 202123037-202123037 on Build GRCh38
rs746692737 Chr.1: 202123114-202123114 on Build GRCh38
rs770548727 Chr.1: 202123119-202123119 on Build GRCh38
rs373409302 Chr.1: 202123090-202123090 on Build GRCh38
rs984354437 Chr.1: 202123035-202123035 on Build GRCh38
rs373957619 Chr.1: 202123221-202123221 on Build GRCh38
rs3795595 Chr.1: 202123232-202123232 on Build GRCh38
rs756909328 Chr.1: 202123192-202123192 on Build GRCh38
rs552502015 Chr.1: 202123169-202123169 on Build GRCh38
rs3795594 Chr.1: 202123204-202123204 on Build GRCh38
rs766487840 Chr.1: 202123257-202123257 on Build GRCh38
rs147299257 Chr.1: 202123254-202123254 on Build GRCh38
rs965126149 Chr.1: 202123214-202123214 on Build GRCh38
rs759964243 Chr.1: 202123049-202123049 on Build GRCh38
rs775080441 Chr.1: 202123206-202123206 on Build GRCh38
rs765717536 Chr.1: 202123051-202123051 on Build GRCh38
rs747974048 Chr.1: 202123102-202123102 on Build GRCh38
rs764564961 Chr.1: 202123155-202123155 on Build GRCh38
rs746759896 Chr.1: 202123180-202123180 on Build GRCh38
rs991318773 Chr.1: 202123200-202123200 on Build GRCh38
rs781163292 Chr.1: 202123075-202123075 on Build GRCh38
rs752647832 Chr.1: 202123171-202123171 on Build GRCh38
rs770243702 Chr.1: 202123128-202123128 on Build GRCh38
rs753878129 Chr.1: 202123263-202123263 on Build GRCh38
rs553879439 Chr.1: 202123099-202123099 on Build GRCh38
rs769333411 Chr.1: 202123205-202123205 on Build GRCh38
rs140390018 Chr.1: 202123129-202123129 on Build GRCh38
rs777578390 Chr.1: 202123175-202123175 on Build GRCh38
rs753061610 Chr.1: 202123064-202123064 on Build GRCh38
rs142876571 Chr.1: 202123158-202123158 on Build GRCh38
rs770447580 Chr.1: 202123045-202123045 on Build GRCh38
rs778901003 Chr.1: 202123091-202123091 on Build GRCh38
rs755255561 Chr.1: 202123169-202123169 on Build GRCh38
rs370435464 Chr.1: 202123157-202123157 on Build GRCh38
rs750453293 Chr.1: 202123089-202123089 on Build GRCh38
rs763406035 Chr.1: 202123068-202123068 on Build GRCh38
rs780866798 Chr.1: 202123198-202123198 on Build GRCh38
rs376395326 Chr.1: 202123135-202123135 on Build GRCh38
rs546366311 Chr.1: 202123073-202123073 on Build GRCh38
rs777492681 Chr.1: 202123112-202123112 on Build GRCh38
rs772817364 Chr.1: 202123041-202123041 on Build GRCh38
rs369524183 Chr.1: 202123069-202123069 on Build GRCh38
rs749815014 Chr.1: 202123210-202123210 on Build GRCh38
rs267598306 Chr.1: 202123034-202123034 on Build GRCh38
rs201014592 Chr.1: 202123048-202123048 on Build GRCh38
rs771829131 Chr.1: 202123108-202123108 on Build GRCh38

View Full Product Details