Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 123577368-123577617 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRAMD1B
Entrez Gene ID
57476
Gene Name
GRAM domain containing 1B
Gene Aliases
-
UniGene
Hs.144725
Gene Chromosome Location
Chr.11: 123358299-123627771 on Build GRCh38
Transcripts
NM_001286563.1 NM_001286564.1 NM_020716.2 XM_005271619.4 XM_005271621.4 XM_005271623.4 XM_005271624.4 XM_006718892.3 XM_011542926.2 XM_011542929.2 XM_011542930.2 XM_011542931.2 XM_017018034.1 XM_017018035.1 XM_017018036.1 XM_017018037.1 XM_017018038.1 XM_017018039.1 XM_017018040.1 XM_017018041.1 XM_017018042.1 XM_017018043.1 XM_017018044.1 XM_017018045.1 XM_017018046.1 XM_017018047.1 XM_017018048.1 XM_017018049.1 XM_017018050.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3444523 c.85C>T p.R29C Chr.11: 123577428-123577428 on Build GRCh38
924176 c.219C>T p.G73G Chr.11: 123577562-123577562 on Build GRCh38
1475131 c.181G>A p.V61M Chr.11: 123577524-123577524 on Build GRCh38
5628011 c.184C>A p.Q62K Chr.11: 123577527-123577527 on Build GRCh38
6025257 c.25A>T p.T9S Chr.11: 123577368-123577368 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777548798 Chr.11: 123577491-123577491 on Build GRCh38
rs760901686 Chr.11: 123577438-123577438 on Build GRCh38
rs775950697 Chr.11: 123577430-123577430 on Build GRCh38
rs76968140 Chr.11: 123577597-123577597 on Build GRCh38
rs750564920 Chr.11: 123577554-123577554 on Build GRCh38
rs576861664 Chr.11: 123577409-123577409 on Build GRCh38
rs894577598 Chr.11: 123577416-123577416 on Build GRCh38
rs764365722 Chr.11: 123577531-123577531 on Build GRCh38
rs1004505378 Chr.11: 123577449-123577449 on Build GRCh38
rs761144627 Chr.11: 123577513-123577513 on Build GRCh38
rs868505983 Chr.11: 123577405-123577405 on Build GRCh38
rs753980128 Chr.11: 123577440-123577440 on Build GRCh38
rs921395364 Chr.11: 123577524-123577524 on Build GRCh38
rs770422939 Chr.11: 123577397-123577397 on Build GRCh38
rs765394893 Chr.11: 123577442-123577442 on Build GRCh38
rs776007839 Chr.11: 123577598-123577598 on Build GRCh38
rs903048868 Chr.11: 123577387-123577387 on Build GRCh38
rs763079105 Chr.11: 123577560-123577560 on Build GRCh38
rs976317856 Chr.11: 123577583-123577583 on Build GRCh38
rs537927876 Chr.11: 123577428-123577428 on Build GRCh38
rs748022861 Chr.11: 123577482-123577482 on Build GRCh38
rs764428433 Chr.11: 123577439-123577439 on Build GRCh38
rs374298813 Chr.11: 123577547-123577547 on Build GRCh38
rs368507512 Chr.11: 123577591-123577591 on Build GRCh38
rs762141069 Chr.11: 123577550-123577550 on Build GRCh38
rs750472988 Chr.11: 123577443-123577443 on Build GRCh38
rs770509019 Chr.11: 123577496-123577496 on Build GRCh38
rs757482018 Chr.11: 123577441-123577441 on Build GRCh38
rs368078644 Chr.11: 123577454-123577454 on Build GRCh38
rs371504166 Chr.11: 123577490-123577490 on Build GRCh38
rs2276409 Chr.11: 123577503-123577503 on Build GRCh38
rs1024315807 Chr.11: 123577447-123577447 on Build GRCh38
rs999972541 Chr.11: 123577398-123577398 on Build GRCh38
rs1054188822 Chr.11: 123577406-123577406 on Build GRCh38
rs745340405 Chr.11: 123577502-123577502 on Build GRCh38
rs773851458 Chr.11: 123577501-123577501 on Build GRCh38
rs765489292 Chr.11: 123577553-123577553 on Build GRCh38
rs760044799 Chr.11: 123577426-123577426 on Build GRCh38
rs574853283 Chr.11: 123577505-123577505 on Build GRCh38
rs1026084418 Chr.11: 123577477-123577477 on Build GRCh38
rs748868785 Chr.11: 123577392-123577392 on Build GRCh38
rs868846413 Chr.11: 123577564-123577564 on Build GRCh38
rs971793495 Chr.11: 123577448-123577448 on Build GRCh38
rs767530052 Chr.11: 123577563-123577563 on Build GRCh38
rs748878338 Chr.11: 123577493-123577493 on Build GRCh38
rs781236429 Chr.11: 123577475-123577475 on Build GRCh38

View Full Product Details