Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 60047198-60047451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEATR6
Entrez Gene ID
63897
Gene Name
HEAT repeat containing 6
Gene Aliases
ABC1
UniGene
Hs.221899
Gene Chromosome Location
Chr.17: 60041363-60078931 on Build GRCh38
Transcripts
NM_022070.4 XM_005257570.3 XM_011525110.2 XM_011525111.2 XM_017024936.1 XM_017024937.1 XM_017024938.1 XM_017024939.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6860715 c.2688A>G p.P896P Chr.17: 60047390-60047390 on Build GRCh38
5628626 c.2680C>A p.P894T Chr.17: 60047398-60047398 on Build GRCh38
6021634 c.2701G>C p.E901Q Chr.17: 60047377-60047377 on Build GRCh38
982138 c.2706C>A p.F902L Chr.17: 60047372-60047372 on Build GRCh38
982136 c.2719C>T p.L907F Chr.17: 60047359-60047359 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757845788 Chr.17: 60047302-60047302 on Build GRCh38
rs567290987 Chr.17: 60047410-60047410 on Build GRCh38
rs114347901 Chr.17: 60047400-60047400 on Build GRCh38
rs34973898 Chr.17: 60047394-60047394 on Build GRCh38
rs371606218 Chr.17: 60047373-60047373 on Build GRCh38
rs1000218984 Chr.17: 60047384-60047384 on Build GRCh38
rs772361062 Chr.17: 60047390-60047390 on Build GRCh38
rs936563079 Chr.17: 60047242-60047242 on Build GRCh38
rs549059644 Chr.17: 60047299-60047299 on Build GRCh38
rs764791267 Chr.17: 60047289-60047289 on Build GRCh38
rs765863341 Chr.17: 60047421-60047421 on Build GRCh38
rs142050052 Chr.17: 60047335-60047335 on Build GRCh38
rs1030924950 Chr.17: 60047386-60047386 on Build GRCh38
rs148810155 Chr.17: 60047322-60047322 on Build GRCh38
rs762481328 Chr.17: 60047412-60047412 on Build GRCh38
rs764879094 Chr.17: 60047404-60047404 on Build GRCh38
rs771072251 Chr.17: 60047363-60047363 on Build GRCh38
rs995466740 Chr.17: 60047392-60047392 on Build GRCh38
rs772784818 Chr.17: 60047408-60047408 on Build GRCh38
rs749396947 Chr.17: 60047348-60047348 on Build GRCh38
rs1053723017 Chr.17: 60047283-60047283 on Build GRCh38
rs551690028 Chr.17: 60047270-60047270 on Build GRCh38
rs536323229 Chr.17: 60047342-60047342 on Build GRCh38
rs367704007 Chr.17: 60047368-60047368 on Build GRCh38
rs754749211 Chr.17: 60047324-60047324 on Build GRCh38
rs200447086 Chr.17: 60047344-60047344 on Build GRCh38
rs779860458 Chr.17: 60047306-60047306 on Build GRCh38
rs753480659 Chr.17: 60047281-60047281 on Build GRCh38
rs761421575 Chr.17: 60047288-60047288 on Build GRCh38
rs761508063 Chr.17: 60047403-60047403 on Build GRCh38
rs151133736 Chr.17: 60047343-60047343 on Build GRCh38
rs895083985 Chr.17: 60047286-60047286 on Build GRCh38

View Full Product Details