Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 160268398-160268623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT80
Entrez Gene ID
57560
Gene Name
intraflagellar transport 80
Gene Aliases
ATD2, SRTD2, WDR56
UniGene
Hs.478095
Gene Chromosome Location
Chr.3: 160256986-160399532 on Build GRCh38
Transcripts
NM_001190241.1 NM_001190242.1 NM_020800.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7108186 c.2139T>A p.D713E Chr.3: 160268497-160268497 on Build GRCh38
6409512 c.2125A>G p.K709E Chr.3: 160268511-160268511 on Build GRCh38
4843147 c.2188G>A p.E730K Chr.3: 160268448-160268448 on Build GRCh38
6420035 c.2113G>T p.A705S Chr.3: 160268523-160268523 on Build GRCh38
2155317 c.2100G>A p.R700R Chr.3: 160268536-160268536 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764764460 Chr.3: 160268481-160268481 on Build GRCh38
rs147761181 Chr.3: 160268584-160268584 on Build GRCh38
rs751917876 Chr.3: 160268494-160268494 on Build GRCh38
rs201820395 Chr.3: 160268478-160268478 on Build GRCh38
rs760611602 Chr.3: 160268448-160268448 on Build GRCh38
rs16831157 Chr.3: 160268497-160268497 on Build GRCh38
rs746088343 Chr.3: 160268557-160268557 on Build GRCh38
rs370294182 Chr.3: 160268487-160268487 on Build GRCh38
rs763798508 Chr.3: 160268460-160268460 on Build GRCh38
rs137853116 Chr.3: 160268535-160268535 on Build GRCh38
rs886058129 Chr.3: 160268431-160268431 on Build GRCh38
rs969072753 Chr.3: 160268438-160268438 on Build GRCh38
rs977361551 Chr.3: 160268457-160268457 on Build GRCh38
rs766079000 Chr.3: 160268490-160268490 on Build GRCh38
rs754140449 Chr.3: 160268551-160268551 on Build GRCh38
rs777972752 Chr.3: 160268533-160268533 on Build GRCh38
rs781425551 Chr.3: 160268504-160268504 on Build GRCh38
rs754746882 Chr.3: 160268504-160268505 on Build GRCh38
rs749975436 Chr.3: 160268485-160268485 on Build GRCh38
rs767350088 Chr.3: 160268530-160268531 on Build GRCh38
rs901815416 Chr.3: 160268427-160268427 on Build GRCh38
rs375941259 Chr.3: 160268436-160268436 on Build GRCh38
rs997566753 Chr.3: 160268462-160268462 on Build GRCh38
rs748643921 Chr.3: 160268529-160268529 on Build GRCh38
rs980650284 Chr.3: 160268503-160268503 on Build GRCh38
rs756615065 Chr.3: 160268530-160268530 on Build GRCh38
rs774644990 Chr.3: 160268540-160268540 on Build GRCh38
rs373671362 Chr.3: 160268536-160268536 on Build GRCh38
rs931581142 Chr.3: 160268550-160268550 on Build GRCh38
rs776319625 Chr.3: 160268587-160268587 on Build GRCh38
rs943067730 Chr.3: 160268520-160268520 on Build GRCh38
rs551983792 Chr.3: 160268480-160268480 on Build GRCh38

View Full Product Details