Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129469245-129469478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT122
Entrez Gene ID
55764
Gene Name
intraflagellar transport 122
Gene Aliases
CED, CED1, SPG, WDR10, WDR10p, WDR140
UniGene
Hs.655284
Gene Chromosome Location
Chr.3: 129440036-129520507 on Build GRCh38
Transcripts
NM_001280541.1 NM_001280545.1 NM_001280546.1 NM_018262.3 NM_052985.3 NM_052989.2 NM_052990.2 XM_005247609.1 XM_006713689.1 XM_006713691.2 XM_006713692.2 XM_006713695.2 XM_011512972.2 XM_017006817.1 XM_017006818.1 XM_017006819.1 XM_017006820.1 XM_017006821.1 XM_017006822.1 XM_017006823.1 XM_017006824.1 XM_017006825.1 XM_017006826.1 XM_017006827.1 XM_017006828.1 XM_017006829.1 XM_017006830.1 XM_017006831.1 XM_017006832.1 XM_017006833.1 XM_017006834.1 XM_017006835.1 XM_017006836.1 XM_017006838.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4639513 c.910A>G p.I304V Chr.3: 129469358-129469358 on Build GRCh38
1038090 c.902G>A p.R301H Chr.3: 129469350-129469350 on Build GRCh38
5574681 c.919G>C p.V307L Chr.3: 129469367-129469367 on Build GRCh38
6479532 c.944C>T p.S315F Chr.3: 129469392-129469392 on Build GRCh38
1130347 c.927C>G p.D309E Chr.3: 129469375-129469375 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761960888 Chr.3: 129469314-129469314 on Build GRCh38
rs866487310 Chr.3: 129469388-129469388 on Build GRCh38
rs770441242 Chr.3: 129469364-129469364 on Build GRCh38
rs113593468 Chr.3: 129469418-129469418 on Build GRCh38
rs754782458 Chr.3: 129469353-129469353 on Build GRCh38
rs749823738 Chr.3: 129469392-129469392 on Build GRCh38
rs185511420 Chr.3: 129469347-129469347 on Build GRCh38
rs376260287 Chr.3: 129469432-129469432 on Build GRCh38
rs766600664 Chr.3: 129469444-129469444 on Build GRCh38
rs774590709 Chr.3: 129469307-129469307 on Build GRCh38
rs746580177 Chr.3: 129469363-129469363 on Build GRCh38
rs368938355 Chr.3: 129469301-129469301 on Build GRCh38
rs752373667 Chr.3: 129469358-129469358 on Build GRCh38
rs373212863 Chr.3: 129469349-129469349 on Build GRCh38
rs778597825 Chr.3: 129469357-129469357 on Build GRCh38
rs772246533 Chr.3: 129469431-129469431 on Build GRCh38
rs748368024 Chr.3: 129469420-129469420 on Build GRCh38
rs372983894 Chr.3: 129469401-129469401 on Build GRCh38
rs976213363 Chr.3: 129469281-129469281 on Build GRCh38
rs970466996 Chr.3: 129469322-129469322 on Build GRCh38
rs143145156 Chr.3: 129469350-129469350 on Build GRCh38
rs758028553 Chr.3: 129469360-129469360 on Build GRCh38
rs202032451 Chr.3: 129469441-129469441 on Build GRCh38
rs769014449 Chr.3: 129469399-129469399 on Build GRCh38
rs750506005 Chr.3: 129469337-129469337 on Build GRCh38
rs758591719 Chr.3: 129469292-129469292 on Build GRCh38
rs1024705335 Chr.3: 129469293-129469293 on Build GRCh38
rs935305563 Chr.3: 129469389-129469389 on Build GRCh38
rs144727222 Chr.3: 129469384-129469384 on Build GRCh38
rs767609724 Chr.3: 129469332-129469332 on Build GRCh38
rs777333238 Chr.3: 129469361-129469361 on Build GRCh38

View Full Product Details