Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 201855718-201855985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IPO9
Entrez Gene ID
55705
Gene Name
importin 9
Gene Aliases
Imp9
UniGene
Hs.596014
Gene Chromosome Location
Chr.1: 201829160-201884294 on Build GRCh38
Transcripts
NM_018085.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4456919 c.1027C>T p.L343L Chr.1: 201855839-201855839 on Build GRCh38
4953882 c.1093A>G p.I365V Chr.1: 201855905-201855905 on Build GRCh38
6287926 c.1083T>G p.I361M Chr.1: 201855895-201855895 on Build GRCh38
4602009 c.1090A>G p.I364V Chr.1: 201855902-201855902 on Build GRCh38
902237 c.1025C>T p.A342V Chr.1: 201855837-201855837 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747275177 Chr.1: 201855780-201855780 on Build GRCh38
rs371470721 Chr.1: 201855752-201855752 on Build GRCh38
rs748120546 Chr.1: 201855928-201855928 on Build GRCh38
rs575695233 Chr.1: 201855837-201855837 on Build GRCh38
rs967333224 Chr.1: 201855765-201855765 on Build GRCh38
rs757562956 Chr.1: 201855897-201855897 on Build GRCh38
rs375198333 Chr.1: 201855814-201855814 on Build GRCh38
rs772836297 Chr.1: 201855839-201855839 on Build GRCh38
rs760390392 Chr.1: 201855852-201855852 on Build GRCh38
rs778210172 Chr.1: 201855894-201855894 on Build GRCh38
rs958275745 Chr.1: 201855864-201855864 on Build GRCh38
rs780244370 Chr.1: 201855920-201855920 on Build GRCh38
rs776772852 Chr.1: 201855804-201855804 on Build GRCh38
rs773065420 Chr.1: 201855939-201855939 on Build GRCh38
rs759024575 Chr.1: 201855868-201855868 on Build GRCh38
rs771848217 Chr.1: 201855935-201855935 on Build GRCh38
rs769965849 Chr.1: 201855919-201855919 on Build GRCh38
rs113909351 Chr.1: 201855796-201855796 on Build GRCh38
rs141794203 Chr.1: 201855809-201855809 on Build GRCh38
rs746432977 Chr.1: 201855938-201855940 on Build GRCh38
rs201318432 Chr.1: 201855751-201855751 on Build GRCh38
rs202009174 Chr.1: 201855879-201855879 on Build GRCh38
rs770997912 Chr.1: 201855791-201855791 on Build GRCh38
rs777957772 Chr.1: 201855777-201855777 on Build GRCh38
rs369226762 Chr.1: 201855895-201855895 on Build GRCh38
rs755481204 Chr.1: 201855759-201855759 on Build GRCh38
rs11576601 Chr.1: 201855824-201855824 on Build GRCh38
rs753454815 Chr.1: 201855866-201855866 on Build GRCh38
rs781261432 Chr.1: 201855907-201855907 on Build GRCh38
rs11578590 Chr.1: 201855823-201855823 on Build GRCh38
rs761742745 Chr.1: 201855834-201855834 on Build GRCh38
rs778839018 Chr.1: 201855872-201855874 on Build GRCh38
rs765881694 Chr.1: 201855855-201855855 on Build GRCh38
rs745997712 Chr.1: 201855913-201855913 on Build GRCh38
rs745934355 Chr.1: 201855808-201855808 on Build GRCh38
rs201275740 Chr.1: 201855763-201855763 on Build GRCh38
rs765772025 Chr.1: 201855873-201855873 on Build GRCh38
rs758730243 Chr.1: 201855772-201855772 on Build GRCh38
rs758851046 Chr.1: 201855889-201855889 on Build GRCh38

View Full Product Details