Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 79362752-79363017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITM2A
Entrez Gene ID
9452
Gene Name
integral membrane protein 2A
Gene Aliases
BRICD2A, E25A
UniGene
Hs.17109
Gene Chromosome Location
Chr.X: 79360384-79367552 on Build GRCh38
Transcripts
NM_001171581.1 NM_004867.4

SNP Detail

SNP ID Chromosome Location
rs767144335 Chr.X: 79362994-79362999 on Build GRCh38
rs374247600 Chr.X: 79362923-79362923 on Build GRCh38
rs758783754 Chr.X: 79362984-79362984 on Build GRCh38
rs773425729 Chr.X: 79362897-79362897 on Build GRCh38
rs888059472 Chr.X: 79362833-79362833 on Build GRCh38
rs929441396 Chr.X: 79362798-79362798 on Build GRCh38
rs752152528 Chr.X: 79362901-79362901 on Build GRCh38
rs953113809 Chr.X: 79362826-79362826 on Build GRCh38
rs36103089 Chr.X: 79362946-79362946 on Build GRCh38
rs943629855 Chr.X: 79362852-79362852 on Build GRCh38
rs768636159 Chr.X: 79362894-79362894 on Build GRCh38
rs941818575 Chr.X: 79362888-79362889 on Build GRCh38
rs758565392 Chr.X: 79362843-79362843 on Build GRCh38
rs200931873 Chr.X: 79362980-79362980 on Build GRCh38
rs377718732 Chr.X: 79362971-79362971 on Build GRCh38
rs369511016 Chr.X: 79362895-79362895 on Build GRCh38
rs967134284 Chr.X: 79362794-79362794 on Build GRCh38
rs764375775 Chr.X: 79362990-79362990 on Build GRCh38
rs760815742 Chr.X: 79362903-79362903 on Build GRCh38
rs144726252 Chr.X: 79362963-79362963 on Build GRCh38
rs919314464 Chr.X: 79362952-79362952 on Build GRCh38
rs752897836 Chr.X: 79362835-79362835 on Build GRCh38
rs776634964 Chr.X: 79362948-79362948 on Build GRCh38
rs779433169 Chr.X: 79362891-79362891 on Build GRCh38
rs922913538 Chr.X: 79362821-79362821 on Build GRCh38
rs774053698 Chr.X: 79362901-79362902 on Build GRCh38
rs763516885 Chr.X: 79362887-79362888 on Build GRCh38
rs749243449 Chr.X: 79362893-79362893 on Build GRCh38
rs761514646 Chr.X: 79362902-79362902 on Build GRCh38
rs781365939 Chr.X: 79362812-79362812 on Build GRCh38
rs768253929 Chr.X: 79362902-79362903 on Build GRCh38

View Full Product Details