Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 19665708-19665977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTR6
Entrez Gene ID
3362
Gene Name
5-hydroxytryptamine receptor 6
Gene Aliases
5-HT6, 5-HT6R
UniGene
Hs.22180
Gene Chromosome Location
Chr.1: 19665287-19680966 on Build GRCh38
Transcripts
NM_000871.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4951982 c.177C>A p.N59K Chr.1: 19665930-19665930 on Build GRCh38
2126040 c.114G>A p.A38A Chr.1: 19665867-19665867 on Build GRCh38
3750737 c.126G>T p.A42A Chr.1: 19665879-19665879 on Build GRCh38
1337684 c.44delG p.A17fs*23 Chr.1: 19665797-19665797 on Build GRCh38
7151247 c.201G>A p.S67S Chr.1: 19665954-19665954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373655326 Chr.1: 19665846-19665846 on Build GRCh38
rs8192529 Chr.1: 19665879-19665879 on Build GRCh38
rs199624661 Chr.1: 19665859-19665859 on Build GRCh38
rs61733141 Chr.1: 19665897-19665897 on Build GRCh38
rs758622864 Chr.1: 19665943-19665943 on Build GRCh38
rs774091625 Chr.1: 19665821-19665822 on Build GRCh38
rs764075775 Chr.1: 19665906-19665906 on Build GRCh38
rs780324617 Chr.1: 19665952-19665952 on Build GRCh38
rs752622966 Chr.1: 19665798-19665798 on Build GRCh38
rs759235818 Chr.1: 19665747-19665747 on Build GRCh38
rs746221490 Chr.1: 19665802-19665802 on Build GRCh38
rs139083731 Chr.1: 19665956-19665956 on Build GRCh38
rs200130105 Chr.1: 19665921-19665921 on Build GRCh38
rs774000123 Chr.1: 19665893-19665893 on Build GRCh38
rs759516289 Chr.1: 19665852-19665853 on Build GRCh38
rs779291764 Chr.1: 19665877-19665877 on Build GRCh38
rs370133428 Chr.1: 19665888-19665888 on Build GRCh38
rs759339647 Chr.1: 19665853-19665853 on Build GRCh38
rs775321136 Chr.1: 19665900-19665900 on Build GRCh38
rs925210549 Chr.1: 19665780-19665780 on Build GRCh38
rs767276826 Chr.1: 19665854-19665854 on Build GRCh38
rs142960547 Chr.1: 19665881-19665881 on Build GRCh38
rs771788727 Chr.1: 19665899-19665899 on Build GRCh38
rs757607144 Chr.1: 19665873-19665873 on Build GRCh38
rs753766815 Chr.1: 19665910-19665910 on Build GRCh38
rs200549575 Chr.1: 19665736-19665736 on Build GRCh38
rs200823780 Chr.1: 19665839-19665839 on Build GRCh38
rs764439006 Chr.1: 19665936-19665936 on Build GRCh38
rs898548076 Chr.1: 19665884-19665884 on Build GRCh38
rs774194160 Chr.1: 19665842-19665842 on Build GRCh38
rs775862853 Chr.1: 19665809-19665809 on Build GRCh38
rs747507674 Chr.1: 19665810-19665810 on Build GRCh38
rs746415893 Chr.1: 19665745-19665745 on Build GRCh38
rs764734222 Chr.1: 19665760-19665760 on Build GRCh38
rs201330629 Chr.1: 19665743-19665743 on Build GRCh38
rs754978772 Chr.1: 19665778-19665778 on Build GRCh38
rs201464687 Chr.1: 19665759-19665759 on Build GRCh38
rs749461476 Chr.1: 19665741-19665741 on Build GRCh38
rs201260093 Chr.1: 19665775-19665775 on Build GRCh38
rs764118329 Chr.1: 19665864-19665864 on Build GRCh38
rs200597696 Chr.1: 19665938-19665938 on Build GRCh38
rs201484895 Chr.1: 19665805-19665805 on Build GRCh38
rs779486915 Chr.1: 19665801-19665801 on Build GRCh38
rs975339306 Chr.1: 19665761-19665761 on Build GRCh38
rs748176940 Chr.1: 19665735-19665735 on Build GRCh38
rs558260074 Chr.1: 19665860-19665860 on Build GRCh38
rs762619989 Chr.1: 19665776-19665776 on Build GRCh38
rs776496438 Chr.1: 19665737-19665737 on Build GRCh38
rs749519244 Chr.1: 19665800-19665800 on Build GRCh38
rs866311551 Chr.1: 19665785-19665785 on Build GRCh38
rs753944730 Chr.1: 19665867-19665867 on Build GRCh38
rs767429118 Chr.1: 19665792-19665792 on Build GRCh38
rs772759671 Chr.1: 19665832-19665832 on Build GRCh38
rs761988087 Chr.1: 19665916-19665916 on Build GRCh38
rs572194723 Chr.1: 19665777-19665777 on Build GRCh38
rs772768813 Chr.1: 19665769-19665769 on Build GRCh38
rs780415156 Chr.1: 19665886-19665886 on Build GRCh38
rs937522837 Chr.1: 19665876-19665876 on Build GRCh38
rs553644433 Chr.1: 19665744-19665744 on Build GRCh38
rs955263858 Chr.1: 19665754-19665754 on Build GRCh38
rs142660187 Chr.1: 19665887-19665887 on Build GRCh38
rs770334471 Chr.1: 19665824-19665824 on Build GRCh38
rs760593809 Chr.1: 19665903-19665903 on Build GRCh38
rs781292195 Chr.1: 19665727-19665727 on Build GRCh38
rs756153475 Chr.1: 19665799-19665799 on Build GRCh38
rs770374013 Chr.1: 19665890-19665890 on Build GRCh38
rs202028782 Chr.1: 19665858-19665858 on Build GRCh38

View Full Product Details