Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 153583471-153583744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0922
Entrez Gene ID
23240
Gene Name
KIAA0922
Gene Aliases
TMEM131L
UniGene
Hs.732450
Gene Chromosome Location
Chr.4: 153466346-153637608 on Build GRCh38
Transcripts
NM_001131007.1 NM_015196.3 XM_011531780.1 XM_011531781.1 XM_011531783.1 XM_017007924.1 XM_017007925.1 XM_017007926.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6209819 c.1004T>A p.V335E Chr.4: 153583616-153583616 on Build GRCh38
377257 c.575C>G p.S192C Chr.4: 153583631-153583631 on Build GRCh38
6209820 c.560T>A p.V187E Chr.4: 153583616-153583616 on Build GRCh38
272854 c.515G>A p.R172H Chr.4: 153583571-153583571 on Build GRCh38
1566908 c.586T>G p.F196V Chr.4: 153583642-153583642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375173085 Chr.4: 153583574-153583574 on Build GRCh38
rs776822840 Chr.4: 153583519-153583519 on Build GRCh38
rs375212049 Chr.4: 153583520-153583520 on Build GRCh38
rs35565709 Chr.4: 153583588-153583588 on Build GRCh38
rs757989948 Chr.4: 153583549-153583550 on Build GRCh38
rs765749687 Chr.4: 153583527-153583527 on Build GRCh38
rs371612098 Chr.4: 153583697-153583697 on Build GRCh38
rs201647921 Chr.4: 153583524-153583524 on Build GRCh38
rs755933991 Chr.4: 153583546-153583546 on Build GRCh38
rs779531792 Chr.4: 153583555-153583557 on Build GRCh38
rs760681541 Chr.4: 153583553-153583553 on Build GRCh38
rs758006990 Chr.4: 153583711-153583711 on Build GRCh38
rs777175655 Chr.4: 153583604-153583604 on Build GRCh38
rs552866877 Chr.4: 153583505-153583505 on Build GRCh38
rs774251099 Chr.4: 153583691-153583691 on Build GRCh38
rs747319418 Chr.4: 153583688-153583688 on Build GRCh38
rs746691251 Chr.4: 153583716-153583716 on Build GRCh38
rs142668158 Chr.4: 153583570-153583570 on Build GRCh38
rs751668900 Chr.4: 153583544-153583544 on Build GRCh38
rs779780455 Chr.4: 153583712-153583712 on Build GRCh38
rs891288554 Chr.4: 153583670-153583670 on Build GRCh38
rs750032237 Chr.4: 153583534-153583535 on Build GRCh38
rs556378667 Chr.4: 153583596-153583596 on Build GRCh38
rs750223385 Chr.4: 153583709-153583709 on Build GRCh38
rs925969482 Chr.4: 153583613-153583613 on Build GRCh38
rs768996966 Chr.4: 153583600-153583600 on Build GRCh38
rs767540792 Chr.4: 153583695-153583695 on Build GRCh38
rs750038344 Chr.4: 153583703-153583703 on Build GRCh38
rs775624626 Chr.4: 153583536-153583536 on Build GRCh38
rs891868166 Chr.4: 153583676-153583676 on Build GRCh38
rs773774362 Chr.4: 153583628-153583628 on Build GRCh38
rs568751420 Chr.4: 153583694-153583694 on Build GRCh38
rs778718135 Chr.4: 153583580-153583580 on Build GRCh38
rs748708879 Chr.4: 153583612-153583612 on Build GRCh38
rs370081967 Chr.4: 153583534-153583534 on Build GRCh38
rs779365116 Chr.4: 153583593-153583593 on Build GRCh38
rs367812835 Chr.4: 153583586-153583586 on Build GRCh38
rs757251127 Chr.4: 153583700-153583700 on Build GRCh38
rs757022111 Chr.4: 153583577-153583577 on Build GRCh38
rs367683043 Chr.4: 153583625-153583626 on Build GRCh38
rs139673482 Chr.4: 153583627-153583627 on Build GRCh38
rs763213912 Chr.4: 153583633-153583633 on Build GRCh38
rs1025374295 Chr.4: 153583510-153583510 on Build GRCh38
rs750644854 Chr.4: 153583529-153583529 on Build GRCh38
rs149777559 Chr.4: 153583653-153583653 on Build GRCh38
rs764804752 Chr.4: 153583640-153583640 on Build GRCh38

View Full Product Details