Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 226794964-226795225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRS1
Entrez Gene ID
3667
Gene Name
insulin receptor substrate 1
Gene Aliases
HIRS-1
UniGene
Hs.471508
Gene Chromosome Location
Chr.2: 226731317-226798790 on Build GRCh38
Transcripts
NM_005544.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6917135 c.3625C>G p.Q1209E Chr.2: 226795114-226795114 on Build GRCh38
1210918 c.3683C>T p.A1228V Chr.2: 226795056-226795056 on Build GRCh38
6687387 c.3658C>T p.R1220C Chr.2: 226795081-226795081 on Build GRCh38
6954946 c.3623A>G p.H1208R Chr.2: 226795116-226795116 on Build GRCh38
6476760 c.3715G>A p.E1239K Chr.2: 226795024-226795024 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747664696 Chr.2: 226795086-226795124 on Build GRCh38
rs147246870 Chr.2: 226795031-226795031 on Build GRCh38
rs750610264 Chr.2: 226794988-226794988 on Build GRCh38
rs774321419 Chr.2: 226795184-226795184 on Build GRCh38
rs777099124 Chr.2: 226795084-226795084 on Build GRCh38
rs747114219 Chr.2: 226795185-226795185 on Build GRCh38
rs376686666 Chr.2: 226795152-226795152 on Build GRCh38
rs548956412 Chr.2: 226795099-226795099 on Build GRCh38
rs762737282 Chr.2: 226795107-226795107 on Build GRCh38
rs150914729 Chr.2: 226795051-226795051 on Build GRCh38
rs375298204 Chr.2: 226795055-226795055 on Build GRCh38
rs780220562 Chr.2: 226794995-226794995 on Build GRCh38
rs1022427396 Chr.2: 226795018-226795018 on Build GRCh38
rs747703859 Chr.2: 226795030-226795030 on Build GRCh38
rs773054335 Chr.2: 226795118-226795119 on Build GRCh38
rs751080359 Chr.2: 226795142-226795142 on Build GRCh38
rs367797493 Chr.2: 226795017-226795017 on Build GRCh38
rs775577355 Chr.2: 226795106-226795106 on Build GRCh38
rs760756636 Chr.2: 226795074-226795074 on Build GRCh38
rs769512509 Chr.2: 226795118-226795147 on Build GRCh38
rs759374843 Chr.2: 226795048-226795048 on Build GRCh38
rs45518734 Chr.2: 226795133-226795133 on Build GRCh38
rs746708337 Chr.2: 226795087-226795087 on Build GRCh38
rs200306802 Chr.2: 226795100-226795100 on Build GRCh38
rs759960668 Chr.2: 226795129-226795129 on Build GRCh38
rs779453938 Chr.2: 226795080-226795080 on Build GRCh38
rs755311653 Chr.2: 226795079-226795079 on Build GRCh38
rs372040825 Chr.2: 226795143-226795143 on Build GRCh38
rs201158268 Chr.2: 226795081-226795081 on Build GRCh38
rs774325390 Chr.2: 226795130-226795131 on Build GRCh38
rs140904930 Chr.2: 226795160-226795160 on Build GRCh38
rs749276374 Chr.2: 226795166-226795166 on Build GRCh38
rs200439296 Chr.2: 226795083-226795083 on Build GRCh38
rs746585066 Chr.2: 226795043-226795043 on Build GRCh38
rs370788917 Chr.2: 226795046-226795046 on Build GRCh38
rs764633017 Chr.2: 226795137-226795137 on Build GRCh38
rs201498527 Chr.2: 226795125-226795125 on Build GRCh38
rs773124039 Chr.2: 226795116-226795116 on Build GRCh38
rs745450452 Chr.2: 226795159-226795159 on Build GRCh38
rs770672949 Chr.2: 226795124-226795125 on Build GRCh38
rs756502280 Chr.2: 226794994-226794994 on Build GRCh38
rs961243937 Chr.2: 226795112-226795112 on Build GRCh38
rs865848090 Chr.2: 226795013-226795013 on Build GRCh38
rs766758551 Chr.2: 226795077-226795077 on Build GRCh38
rs772212335 Chr.2: 226795115-226795115 on Build GRCh38
rs777548680 Chr.2: 226795035-226795035 on Build GRCh38
rs547405267 Chr.2: 226795005-226795005 on Build GRCh38
rs779660430 Chr.2: 226795165-226795165 on Build GRCh38
rs973649983 Chr.2: 226795104-226795104 on Build GRCh38
rs537194227 Chr.2: 226795094-226795094 on Build GRCh38
rs770781927 Chr.2: 226795045-226795045 on Build GRCh38
rs769904403 Chr.2: 226795049-226795049 on Build GRCh38
rs199818392 Chr.2: 226795177-226795177 on Build GRCh38
rs1042439056 Chr.2: 226795063-226795063 on Build GRCh38
rs765518355 Chr.2: 226795130-226795130 on Build GRCh38
rs760773926 Chr.2: 226795118-226795118 on Build GRCh38
rs766635973 Chr.2: 226795124-226795124 on Build GRCh38
rs750668066 Chr.2: 226795071-226795071 on Build GRCh38
rs754239320 Chr.2: 226795078-226795078 on Build GRCh38
rs557233754 Chr.2: 226795069-226795069 on Build GRCh38
rs753270746 Chr.2: 226795131-226795131 on Build GRCh38
rs150039193 Chr.2: 226795057-226795057 on Build GRCh38

View Full Product Details