Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 163471260-163471499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMMR
Entrez Gene ID
3161
Gene Name
hyaluronan mediated motility receptor
Gene Aliases
CD168, IHABP, RHAMM
UniGene
Hs.740467
Gene Chromosome Location
Chr.5: 163460511-163491945 on Build GRCh38
Transcripts
NM_001142556.1 NM_001142557.1 NM_012484.2 NM_012485.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4959098 c.620G>A p.G207E Chr.5: 163471436-163471436 on Build GRCh38
3919546 c.564A>C p.Q188H Chr.5: 163471380-163471380 on Build GRCh38
5975494 c.568G>A p.G190S Chr.5: 163471384-163471384 on Build GRCh38
1065371 c.553A>G p.M185V Chr.5: 163471369-163471369 on Build GRCh38
1065372 c.607G>A p.E203K Chr.5: 163471423-163471423 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752662707 Chr.5: 163471388-163471388 on Build GRCh38
rs199572983 Chr.5: 163471333-163471333 on Build GRCh38
rs758493057 Chr.5: 163471420-163471420 on Build GRCh38
rs535193481 Chr.5: 163471383-163471383 on Build GRCh38
rs755354257 Chr.5: 163471433-163471433 on Build GRCh38
rs547199953 Chr.5: 163471393-163471393 on Build GRCh38
rs763139642 Chr.5: 163471464-163471465 on Build GRCh38
rs748979552 Chr.5: 163471452-163471452 on Build GRCh38
rs772891277 Chr.5: 163471296-163471296 on Build GRCh38
rs955551879 Chr.5: 163471366-163471366 on Build GRCh38
rs770777678 Chr.5: 163471314-163471314 on Build GRCh38
rs762472075 Chr.5: 163471301-163471301 on Build GRCh38
rs776787151 Chr.5: 163471302-163471303 on Build GRCh38
rs144523729 Chr.5: 163471456-163471456 on Build GRCh38
rs762028633 Chr.5: 163471389-163471389 on Build GRCh38
rs201965952 Chr.5: 163471423-163471423 on Build GRCh38
rs376665983 Chr.5: 163471422-163471422 on Build GRCh38
rs769269790 Chr.5: 163471295-163471295 on Build GRCh38
rs759520244 Chr.5: 163471368-163471368 on Build GRCh38
rs750680641 Chr.5: 163471418-163471418 on Build GRCh38
rs115147384 Chr.5: 163471281-163471281 on Build GRCh38
rs775729353 Chr.5: 163471464-163471467 on Build GRCh38
rs759672327 Chr.5: 163471396-163471397 on Build GRCh38
rs765297224 Chr.5: 163471416-163471431 on Build GRCh38
rs777211342 Chr.5: 163471448-163471448 on Build GRCh38
rs748622236 Chr.5: 163471440-163471440 on Build GRCh38
rs770419663 Chr.5: 163471455-163471455 on Build GRCh38
rs888200990 Chr.5: 163471319-163471319 on Build GRCh38
rs781700042 Chr.5: 163471434-163471434 on Build GRCh38

View Full Product Details