Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45149015-45149274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEXIM1
Entrez Gene ID
10614
Gene Name
hexamethylene bisacetamide inducible 1
Gene Aliases
CLP1, EDG1, HIS1, MAQ1
UniGene
Hs.586945
Gene Chromosome Location
Chr.17: 45147317-45152101 on Build GRCh38
Transcripts
NM_006460.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
71053 c.49A>T p.N17Y Chr.17: 45149239-45149239 on Build GRCh38
472902 c.55A>C p.T19P Chr.17: 45149245-45149245 on Build GRCh38
1662131 c.76G>C p.E26Q Chr.17: 45149266-45149266 on Build GRCh38
7174088 c.77A>T p.E26V Chr.17: 45149267-45149267 on Build GRCh38
4634203 c.11C>A p.P4Q Chr.17: 45149201-45149201 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs67016906 Chr.17: 45149111-45149111 on Build GRCh38
rs371225086 Chr.17: 45149181-45149181 on Build GRCh38
rs765719338 Chr.17: 45149216-45149216 on Build GRCh38
rs78652521 Chr.17: 45149095-45149095 on Build GRCh38
rs7216041 Chr.17: 45149111-45149111 on Build GRCh38
rs77862216 Chr.17: 45149094-45149094 on Build GRCh38
rs763523915 Chr.17: 45149184-45149184 on Build GRCh38
rs779309521 Chr.17: 45149111-45149112 on Build GRCh38
rs142925080 Chr.17: 45149052-45149052 on Build GRCh38
rs745618309 Chr.17: 45149155-45149155 on Build GRCh38
rs529783521 Chr.17: 45149140-45149140 on Build GRCh38
rs778014257 Chr.17: 45149142-45149142 on Build GRCh38
rs200065196 Chr.17: 45149167-45149167 on Build GRCh38
rs759056628 Chr.17: 45149176-45149186 on Build GRCh38
rs958723875 Chr.17: 45149079-45149079 on Build GRCh38
rs373987297 Chr.17: 45149150-45149150 on Build GRCh38
rs111687345 Chr.17: 45149105-45149106 on Build GRCh38
rs1017604362 Chr.17: 45149104-45149105 on Build GRCh38
rs543354181 Chr.17: 45149086-45149086 on Build GRCh38
rs776678432 Chr.17: 45149154-45149154 on Build GRCh38
rs967844637 Chr.17: 45149139-45149139 on Build GRCh38
rs7216036 Chr.17: 45149105-45149105 on Build GRCh38
rs892302792 Chr.17: 45149080-45149080 on Build GRCh38
rs936157277 Chr.17: 45149171-45149171 on Build GRCh38
rs774202797 Chr.17: 45149143-45149158 on Build GRCh38
rs766775697 Chr.17: 45149199-45149199 on Build GRCh38
rs367729540 Chr.17: 45149249-45149249 on Build GRCh38
rs773843252 Chr.17: 45149190-45149190 on Build GRCh38
rs778210451 Chr.17: 45149231-45149231 on Build GRCh38
rs538964222 Chr.17: 45149100-45149100 on Build GRCh38
rs903936584 Chr.17: 45149103-45149105 on Build GRCh38
rs1056751228 Chr.17: 45149067-45149067 on Build GRCh38
rs993265070 Chr.17: 45149099-45149099 on Build GRCh38
rs752910152 Chr.17: 45149222-45149222 on Build GRCh38
rs755261268 Chr.17: 45149203-45149203 on Build GRCh38
rs758700108 Chr.17: 45149228-45149228 on Build GRCh38
rs140416514 Chr.17: 45149255-45149255 on Build GRCh38
rs747061601 Chr.17: 45149236-45149236 on Build GRCh38
rs770771089 Chr.17: 45149153-45149153 on Build GRCh38
rs372493500 Chr.17: 45149105-45149105 on Build GRCh38
rs369455400 Chr.17: 45149110-45149111 on Build GRCh38
rs7220808 Chr.17: 45149104-45149104 on Build GRCh38
rs754411480 Chr.17: 45149201-45149201 on Build GRCh38
rs1034266815 Chr.17: 45149060-45149060 on Build GRCh38
rs559367587 Chr.17: 45149123-45149123 on Build GRCh38
rs373817429 Chr.17: 45149237-45149237 on Build GRCh38
rs761441239 Chr.17: 45149195-45149195 on Build GRCh38
rs1009333138 Chr.17: 45149100-45149105 on Build GRCh38
rs769660725 Chr.17: 45149165-45149165 on Build GRCh38
rs375433684 Chr.17: 45149108-45149111 on Build GRCh38

View Full Product Details