Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101064823-101065048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KAZALD1
Entrez Gene ID
81621
Gene Name
Kazal type serine peptidase inhibitor domain 1
Gene Aliases
BONO1, FKSG28, FKSG40, IGFBP-rP10
UniGene
Hs.534859
Gene Chromosome Location
Chr.10: 101061141-101068141 on Build GRCh38
Transcripts
NM_001319303.1 NM_030929.4 NR_135067.1 XM_017016714.1 XM_017016715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5913054 c.874G>T p.E292* Chr.10: 101064879-101064879 on Build GRCh38
914021 c.839G>A p.G280D Chr.10: 101064844-101064844 on Build GRCh38
4621763 c.845C>T p.P282L Chr.10: 101064850-101064850 on Build GRCh38
1345380 c.904G>A p.D302N Chr.10: 101064909-101064909 on Build GRCh38
7002840 c.862A>T p.N288Y Chr.10: 101064867-101064867 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs183547624 Chr.10: 101064909-101064909 on Build GRCh38
rs761195884 Chr.10: 101064873-101064873 on Build GRCh38
rs762436641 Chr.10: 101064882-101064882 on Build GRCh38
rs370620721 Chr.10: 101064935-101064935 on Build GRCh38
rs755106497 Chr.10: 101064932-101064932 on Build GRCh38
rs772415022 Chr.10: 101064869-101064869 on Build GRCh38
rs1013225273 Chr.10: 101064862-101064862 on Build GRCh38
rs11818875 Chr.10: 101065017-101065017 on Build GRCh38
rs754034117 Chr.10: 101064924-101064924 on Build GRCh38
rs200601082 Chr.10: 101064938-101064938 on Build GRCh38
rs570988991 Chr.10: 101065014-101065014 on Build GRCh38
rs181846212 Chr.10: 101065016-101065016 on Build GRCh38
rs761523575 Chr.10: 101064891-101064891 on Build GRCh38
rs773181682 Chr.10: 101064891-101064892 on Build GRCh38
rs761482148 Chr.10: 101064970-101064970 on Build GRCh38
rs778160871 Chr.10: 101064952-101064952 on Build GRCh38
rs192613188 Chr.10: 101064858-101064858 on Build GRCh38
rs763801399 Chr.10: 101064884-101064884 on Build GRCh38
rs754637769 Chr.10: 101064936-101064936 on Build GRCh38
rs767291203 Chr.10: 101064908-101064908 on Build GRCh38
rs779285694 Chr.10: 101064937-101064937 on Build GRCh38
rs368900664 Chr.10: 101064859-101064859 on Build GRCh38
rs371220007 Chr.10: 101064923-101064923 on Build GRCh38
rs760144303 Chr.10: 101064904-101064909 on Build GRCh38
rs1017598219 Chr.10: 101064980-101064980 on Build GRCh38
rs773895183 Chr.10: 101064962-101064962 on Build GRCh38
rs374997226 Chr.10: 101064953-101064953 on Build GRCh38
rs773607207 Chr.10: 101064872-101064872 on Build GRCh38
rs563015684 Chr.10: 101064874-101064874 on Build GRCh38
rs531946008 Chr.10: 101065015-101065015 on Build GRCh38
rs189458098 Chr.10: 101064999-101064999 on Build GRCh38
rs768093857 Chr.10: 101064961-101064961 on Build GRCh38
rs775029615 Chr.10: 101064877-101064877 on Build GRCh38
rs769052567 Chr.10: 101064850-101064850 on Build GRCh38
rs994368861 Chr.10: 101064916-101064916 on Build GRCh38
rs530015590 Chr.10: 101064887-101064887 on Build GRCh38
rs772345105 Chr.10: 101064946-101064946 on Build GRCh38
rs771485739 Chr.10: 101064954-101064954 on Build GRCh38
rs989692104 Chr.10: 101064928-101064928 on Build GRCh38
rs927974722 Chr.10: 101064994-101064994 on Build GRCh38
rs762452970 Chr.10: 101064959-101064959 on Build GRCh38
rs767513445 Chr.10: 101064848-101064848 on Build GRCh38
rs778058529 Chr.10: 101064947-101064947 on Build GRCh38
rs777116917 Chr.10: 101064958-101064958 on Build GRCh38
rs1042274528 Chr.10: 101064990-101064990 on Build GRCh38
rs920977802 Chr.10: 101064897-101064897 on Build GRCh38
rs12359843 Chr.10: 101065003-101065003 on Build GRCh38

View Full Product Details