Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 112978530-112978803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL36G
Entrez Gene ID
56300
Gene Name
interleukin 36, gamma
Gene Aliases
IL-1F9, IL-1H1, IL-1RP2, IL1E, IL1F9, IL1H1, IL1RP2
UniGene
Hs.211238
Gene Chromosome Location
Chr.2: 112978019-112985672 on Build GRCh38
Transcripts
NM_001278568.1 NM_019618.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1690915 c.23C>T p.A8V Chr.2: 112978661-112978661 on Build GRCh38
39629 c.42C>T p.A14A Chr.2: 112978680-112978680 on Build GRCh38
213850 c.43G>C p.V15L Chr.2: 112978681-112978681 on Build GRCh38
3565428 c.49C>T p.Q17* Chr.2: 112978687-112978687 on Build GRCh38
3894551 c.35G>A p.G12E Chr.2: 112978673-112978673 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547422708 Chr.2: 112978708-112978708 on Build GRCh38
rs749349270 Chr.2: 112978733-112978733 on Build GRCh38
rs766704475 Chr.2: 112978589-112978589 on Build GRCh38
rs888935631 Chr.2: 112978759-112978759 on Build GRCh38
rs769970410 Chr.2: 112978658-112978658 on Build GRCh38
rs746220303 Chr.2: 112978742-112978742 on Build GRCh38
rs368856752 Chr.2: 112978680-112978680 on Build GRCh38
rs753322717 Chr.2: 112978625-112978625 on Build GRCh38
rs748815896 Chr.2: 112978678-112978679 on Build GRCh38
rs764786003 Chr.2: 112978630-112978630 on Build GRCh38
rs373779128 Chr.2: 112978720-112978720 on Build GRCh38
rs28947208 Chr.2: 112978758-112978758 on Build GRCh38
rs749393855 Chr.2: 112978752-112978752 on Build GRCh38
rs367945118 Chr.2: 112978737-112978737 on Build GRCh38
rs371482572 Chr.2: 112978598-112978598 on Build GRCh38
rs28947207 Chr.2: 112978748-112978748 on Build GRCh38
rs758024889 Chr.2: 112978703-112978703 on Build GRCh38
rs201477223 Chr.2: 112978710-112978710 on Build GRCh38
rs754884422 Chr.2: 112978711-112978711 on Build GRCh38
rs781315646 Chr.2: 112978640-112978640 on Build GRCh38
rs776035980 Chr.2: 112978676-112978676 on Build GRCh38
rs997344480 Chr.2: 112978577-112978578 on Build GRCh38
rs371389287 Chr.2: 112978582-112978582 on Build GRCh38
rs770045511 Chr.2: 112978741-112978741 on Build GRCh38
rs771089638 Chr.2: 112978660-112978660 on Build GRCh38
rs768032731 Chr.2: 112978623-112978623 on Build GRCh38
rs181260849 Chr.2: 112978688-112978688 on Build GRCh38
rs759995603 Chr.2: 112978620-112978620 on Build GRCh38
rs28947206 Chr.2: 112978719-112978719 on Build GRCh38
rs777867952 Chr.2: 112978721-112978721 on Build GRCh38
rs770413221 Chr.2: 112978746-112978746 on Build GRCh38
rs757717589 Chr.2: 112978605-112978605 on Build GRCh38
rs748230876 Chr.2: 112978645-112978645 on Build GRCh38
rs900139030 Chr.2: 112978749-112978749 on Build GRCh38
rs750034166 Chr.2: 112978631-112978631 on Build GRCh38
rs747478548 Chr.2: 112978570-112978570 on Build GRCh38
rs768009971 Chr.2: 112978674-112978674 on Build GRCh38
rs756860964 Chr.2: 112978629-112978629 on Build GRCh38
rs746826301 Chr.2: 112978638-112978638 on Build GRCh38
rs199830538 Chr.2: 112978633-112978633 on Build GRCh38
rs571391180 Chr.2: 112978659-112978659 on Build GRCh38
rs764738286 Chr.2: 112978681-112978681 on Build GRCh38
rs759940517 Chr.2: 112978662-112978662 on Build GRCh38
rs754946711 Chr.2: 112978639-112978639 on Build GRCh38

View Full Product Details