Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97936130-97936355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEMGN
Entrez Gene ID
55363
Gene Name
hemogen
Gene Aliases
CT155, EDAG, EDAG-1, NDR
UniGene
Hs.176626
Gene Chromosome Location
Chr.9: 97926791-97944915 on Build GRCh38
Transcripts
NM_018437.4 NM_197978.2 XM_005252086.1 XM_011518845.2 XM_011518846.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4996987 c.164G>T p.W55L Chr.9: 97936180-97936180 on Build GRCh38
6901182 c.80-1G>A p.? Chr.9: 97936265-97936265 on Build GRCh38
4998893 c.142C>T p.H48Y Chr.9: 97936202-97936202 on Build GRCh38
1103076 c.110G>T p.R37I Chr.9: 97936234-97936234 on Build GRCh38
1209522 c.128G>T p.R43I Chr.9: 97936216-97936216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757846898 Chr.9: 97936262-97936262 on Build GRCh38
rs775579172 Chr.9: 97936250-97936250 on Build GRCh38
rs762350601 Chr.9: 97936256-97936256 on Build GRCh38
rs774638516 Chr.9: 97936203-97936203 on Build GRCh38
rs200915997 Chr.9: 97936282-97936282 on Build GRCh38
rs368458637 Chr.9: 97936190-97936190 on Build GRCh38
rs562872909 Chr.9: 97936187-97936187 on Build GRCh38
rs576432009 Chr.9: 97936295-97936295 on Build GRCh38
rs751016338 Chr.9: 97936302-97936302 on Build GRCh38
rs754690331 Chr.9: 97936313-97936313 on Build GRCh38
rs781171008 Chr.9: 97936297-97936297 on Build GRCh38
rs151137134 Chr.9: 97936202-97936202 on Build GRCh38
rs184858948 Chr.9: 97936253-97936253 on Build GRCh38
rs565073856 Chr.9: 97936276-97936276 on Build GRCh38
rs188059572 Chr.9: 97936320-97936320 on Build GRCh38
rs368686654 Chr.9: 97936311-97936311 on Build GRCh38
rs866330659 Chr.9: 97936193-97936193 on Build GRCh38
rs756178703 Chr.9: 97936307-97936307 on Build GRCh38
rs764466239 Chr.9: 97936252-97936252 on Build GRCh38
rs759663815 Chr.9: 97936235-97936235 on Build GRCh38
rs373653126 Chr.9: 97936304-97936304 on Build GRCh38
rs758141430 Chr.9: 97936163-97936163 on Build GRCh38
rs58597487 Chr.9: 97936222-97936222 on Build GRCh38
rs746747580 Chr.9: 97936176-97936176 on Build GRCh38
rs745894960 Chr.9: 97936312-97936312 on Build GRCh38
rs146948225 Chr.9: 97936251-97936251 on Build GRCh38
rs948628827 Chr.9: 97936172-97936172 on Build GRCh38
rs764727204 Chr.9: 97936258-97936258 on Build GRCh38
rs750011388 Chr.9: 97936261-97936261 on Build GRCh38
rs772280001 Chr.9: 97936243-97936243 on Build GRCh38
rs751190638 Chr.9: 97936287-97936287 on Build GRCh38
rs766829765 Chr.9: 97936212-97936213 on Build GRCh38
rs748142802 Chr.9: 97936200-97936200 on Build GRCh38
rs754324765 Chr.9: 97936255-97936255 on Build GRCh38
rs779705074 Chr.9: 97936169-97936169 on Build GRCh38
rs781224103 Chr.9: 97936198-97936198 on Build GRCh38

View Full Product Details