Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 35434733-35435004 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0319L
Entrez Gene ID
79932
Gene Name
KIAA0319 like
Gene Aliases
AAVR
UniGene
Hs.456507
Gene Chromosome Location
Chr.1: 35433490-35557670 on Build GRCh38
Transcripts
NM_024874.4 XM_006710907.1 XM_006710909.1 XM_006710910.1 XM_011542179.2 XM_011542180.1 XM_017002366.1 XM_017002367.1 XM_017002368.1 XM_017002369.1 XM_017002370.1 XM_017002372.1 XM_017002374.1 XM_017002375.1 XM_017002376.1 XM_017002377.1 XM_017002378.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5674762 c.3134G>T p.R1045L Chr.1: 35434910-35434910 on Build GRCh38
3488908 c.3111C>T p.T1037T Chr.1: 35434933-35434933 on Build GRCh38
1342140 c.3056G>A p.R1019Q Chr.1: 35434988-35434988 on Build GRCh38
7373484 c.3091T>C p.S1031P Chr.1: 35434953-35434953 on Build GRCh38
4695224 c.3045A>G p.T1015T Chr.1: 35434999-35434999 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746172922 Chr.1: 35434975-35434975 on Build GRCh38
rs375670791 Chr.1: 35434850-35434850 on Build GRCh38
rs934369251 Chr.1: 35434788-35434788 on Build GRCh38
rs779012868 Chr.1: 35434892-35434892 on Build GRCh38
rs374202583 Chr.1: 35434984-35434984 on Build GRCh38
rs762519844 Chr.1: 35434890-35434890 on Build GRCh38
rs200186246 Chr.1: 35434919-35434919 on Build GRCh38
rs187691490 Chr.1: 35434845-35434845 on Build GRCh38
rs925641196 Chr.1: 35434787-35434787 on Build GRCh38
rs944241382 Chr.1: 35434823-35434823 on Build GRCh38
rs200198418 Chr.1: 35434877-35434877 on Build GRCh38
rs1052906180 Chr.1: 35434812-35434812 on Build GRCh38
rs747507850 Chr.1: 35434931-35434931 on Build GRCh38
rs761505736 Chr.1: 35434863-35434863 on Build GRCh38
rs780034518 Chr.1: 35434929-35434929 on Build GRCh38
rs770436309 Chr.1: 35434971-35434971 on Build GRCh38
rs150532754 Chr.1: 35434816-35434816 on Build GRCh38
rs548927392 Chr.1: 35434965-35434965 on Build GRCh38
rs368457639 Chr.1: 35434947-35434947 on Build GRCh38
rs757502304 Chr.1: 35434882-35434883 on Build GRCh38
rs560972010 Chr.1: 35434798-35434798 on Build GRCh38
rs549847818 Chr.1: 35434830-35434830 on Build GRCh38
rs765857225 Chr.1: 35434893-35434893 on Build GRCh38
rs759424858 Chr.1: 35434904-35434904 on Build GRCh38
rs769064246 Chr.1: 35434941-35434941 on Build GRCh38
rs779108241 Chr.1: 35434913-35434913 on Build GRCh38
rs773758200 Chr.1: 35434980-35434980 on Build GRCh38
rs749829544 Chr.1: 35434938-35434938 on Build GRCh38
rs751484493 Chr.1: 35434896-35434896 on Build GRCh38
rs750698281 Chr.1: 35434916-35434916 on Build GRCh38
rs781641046 Chr.1: 35434942-35434942 on Build GRCh38
rs368905748 Chr.1: 35434882-35434882 on Build GRCh38
rs770349013 Chr.1: 35434955-35434955 on Build GRCh38
rs374202036 Chr.1: 35434912-35434912 on Build GRCh38
rs373199789 Chr.1: 35434910-35434910 on Build GRCh38
rs780592964 Chr.1: 35434847-35434847 on Build GRCh38
rs139539582 Chr.1: 35434831-35434831 on Build GRCh38
rs370808394 Chr.1: 35434948-35434948 on Build GRCh38
rs777164957 Chr.1: 35434851-35434851 on Build GRCh38

View Full Product Details