Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 113083889-113084150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR85
Entrez Gene ID
54329
Gene Name
G protein-coupled receptor 85
Gene Aliases
SREB, SREB2
UniGene
Hs.152009
Gene Chromosome Location
Chr.7: 113080413-113087778 on Build GRCh38
Transcripts
NM_001146265.1 NM_001146266.1 NM_001146267.1 NM_018970.6 XM_005250451.2 XM_017012352.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1084151 c.704C>T p.A235V Chr.7: 113084018-113084018 on Build GRCh38
3256419 c.624C>T p.F208F Chr.7: 113084098-113084098 on Build GRCh38
3256415 c.685A>G p.T229A Chr.7: 113084037-113084037 on Build GRCh38
1084153 c.595C>T p.Q199* Chr.7: 113084127-113084127 on Build GRCh38
1084152 c.634C>T p.R212* Chr.7: 113084088-113084088 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187730830 Chr.7: 113084031-113084031 on Build GRCh38
rs751456164 Chr.7: 113084089-113084089 on Build GRCh38
rs779473547 Chr.7: 113084121-113084121 on Build GRCh38
rs770417550 Chr.7: 113083959-113083959 on Build GRCh38
rs750649000 Chr.7: 113084095-113084095 on Build GRCh38
rs780450966 Chr.7: 113084098-113084098 on Build GRCh38
rs138737854 Chr.7: 113083999-113083999 on Build GRCh38
rs779305786 Chr.7: 113083916-113083916 on Build GRCh38
rs767607685 Chr.7: 113083957-113083957 on Build GRCh38
rs746465211 Chr.7: 113083930-113083930 on Build GRCh38
rs764034931 Chr.7: 113084077-113084077 on Build GRCh38
rs920604596 Chr.7: 113083960-113083960 on Build GRCh38
rs112218049 Chr.7: 113083913-113083915 on Build GRCh38
rs996876186 Chr.7: 113083968-113083968 on Build GRCh38
rs973364057 Chr.7: 113083922-113083922 on Build GRCh38
rs755306312 Chr.7: 113084110-113084110 on Build GRCh38
rs917408763 Chr.7: 113084045-113084045 on Build GRCh38
rs770454809 Chr.7: 113084127-113084127 on Build GRCh38
rs1032173836 Chr.7: 113084113-113084113 on Build GRCh38
rs28392728 Chr.7: 113084092-113084092 on Build GRCh38
rs958791679 Chr.7: 113083967-113083967 on Build GRCh38
rs754202003 Chr.7: 113084100-113084100 on Build GRCh38
rs201130668 Chr.7: 113084063-113084063 on Build GRCh38
rs756442417 Chr.7: 113084097-113084097 on Build GRCh38
rs373221793 Chr.7: 113084072-113084072 on Build GRCh38
rs769548475 Chr.7: 113084056-113084056 on Build GRCh38
rs767549363 Chr.7: 113084094-113084094 on Build GRCh38
rs1041947361 Chr.7: 113084091-113084091 on Build GRCh38
rs748663821 Chr.7: 113084125-113084125 on Build GRCh38

View Full Product Details