Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 50592889-50593144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRB10
Entrez Gene ID
2887
Gene Name
growth factor receptor bound protein 10
Gene Aliases
GRB-IR, Grb-10, IRBP, MEG1, RSS
UniGene
Hs.164060
Gene Chromosome Location
Chr.7: 50590063-50793462 on Build GRCh38
Transcripts
NM_001001549.2 NM_001001550.2 NM_001001555.2 NM_005311.4 XM_011515304.1 XM_011515312.2 XM_011515316.1 XM_011515323.2 XM_017012029.1 XM_017012030.1 XM_017012031.1 XM_017012032.1 XM_017012033.1 XM_017012034.1 XM_017012035.1 XM_017012036.1 XM_017012037.1 XM_017012038.1 XM_017012039.1 XM_017012040.1 XM_017012041.1 XM_017012042.1 XM_017012043.1 XM_017012044.1 XM_017012045.1 XM_017012046.1 XM_017012047.1 XM_017012048.1 XM_017012049.1 XM_017012050.1 XM_017012051.1 XM_017012052.1 XM_017012053.1 XM_017012054.1 XM_017012055.1 XM_017012056.1 XM_017012057.1 XM_017012058.1 XM_017012059.1 XM_017012060.1 XM_017012061.1 XM_017012062.1 XM_017012063.1 XM_017012064.1 XM_017012065.1 XM_017012066.1 XM_017012067.1 XM_017012068.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1313166 c.1755C>G p.L585L Chr.7: 50592982-50592982 on Build GRCh38
485436 c.1777G>A p.A593T Chr.7: 50592960-50592960 on Build GRCh38
6350584 c.1629C>T p.D543D Chr.7: 50593090-50593090 on Build GRCh38
1316310 c.1707G>T p.Q569H Chr.7: 50593030-50593030 on Build GRCh38
7317309 c.1675G>A p.D559N Chr.7: 50593062-50593062 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779098154 Chr.7: 50593087-50593087 on Build GRCh38
rs746530062 Chr.7: 50592914-50592914 on Build GRCh38
rs769446325 Chr.7: 50592935-50592935 on Build GRCh38
rs762262034 Chr.7: 50593112-50593112 on Build GRCh38
rs369570284 Chr.7: 50593126-50593126 on Build GRCh38
rs537408631 Chr.7: 50592965-50592965 on Build GRCh38
rs3807550 Chr.7: 50593090-50593090 on Build GRCh38
rs774351478 Chr.7: 50592945-50592945 on Build GRCh38
rs751149581 Chr.7: 50593123-50593123 on Build GRCh38
rs919918053 Chr.7: 50592950-50592950 on Build GRCh38
rs761279838 Chr.7: 50592970-50592970 on Build GRCh38
rs748174069 Chr.7: 50592931-50592931 on Build GRCh38
rs749525996 Chr.7: 50593102-50593103 on Build GRCh38
rs780986977 Chr.7: 50592930-50592930 on Build GRCh38
rs762633448 Chr.7: 50593011-50593011 on Build GRCh38
rs11768472 Chr.7: 50593065-50593065 on Build GRCh38
rs771156851 Chr.7: 50592943-50592943 on Build GRCh38
rs777488599 Chr.7: 50593079-50593079 on Build GRCh38
rs777067081 Chr.7: 50593105-50593105 on Build GRCh38
rs752538444 Chr.7: 50593075-50593075 on Build GRCh38
rs749089190 Chr.7: 50593084-50593084 on Build GRCh38
rs972527779 Chr.7: 50592960-50592960 on Build GRCh38
rs779659227 Chr.7: 50592911-50592911 on Build GRCh38
rs764509734 Chr.7: 50592982-50592982 on Build GRCh38
rs765754062 Chr.7: 50593119-50593119 on Build GRCh38
rs776688540 Chr.7: 50593029-50593029 on Build GRCh38
rs754876636 Chr.7: 50592915-50592915 on Build GRCh38
rs780795259 Chr.7: 50593070-50593070 on Build GRCh38
rs375434973 Chr.7: 50593096-50593096 on Build GRCh38
rs766132484 Chr.7: 50593013-50593013 on Build GRCh38
rs754273501 Chr.7: 50592985-50592985 on Build GRCh38
rs775666513 Chr.7: 50593095-50593095 on Build GRCh38
rs775842626 Chr.7: 50592966-50592966 on Build GRCh38
rs751290362 Chr.7: 50593060-50593060 on Build GRCh38
rs576031631 Chr.7: 50592936-50592936 on Build GRCh38
rs369791487 Chr.7: 50593086-50593086 on Build GRCh38
rs759533052 Chr.7: 50592949-50592949 on Build GRCh38
rs769297711 Chr.7: 50593104-50593104 on Build GRCh38
rs746154687 Chr.7: 50593089-50593089 on Build GRCh38
rs754432446 Chr.7: 50593066-50593066 on Build GRCh38
rs375224737 Chr.7: 50593078-50593078 on Build GRCh38
rs960610076 Chr.7: 50593027-50593027 on Build GRCh38
rs768819777 Chr.7: 50593114-50593114 on Build GRCh38
rs372347272 Chr.7: 50592941-50592941 on Build GRCh38

View Full Product Details