Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 21304597-21304846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFNA5
Entrez Gene ID
3442
Gene Name
interferon alpha 5
Gene Aliases
IFN-alpha-5, IFN-alphaG, INA5, INFA5, leIF G
UniGene
Hs.37113
Gene Chromosome Location
Chr.9: 21304614-21305313 on Build GRCh38
Transcripts
NM_002169.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6204698 c.470A>G p.K157R Chr.9: 21304787-21304787 on Build GRCh38
4588739 c.416C>T p.S139F Chr.9: 21304841-21304841 on Build GRCh38
7149346 c.508G>C p.E170Q Chr.9: 21304749-21304749 on Build GRCh38
4827516 c.442C>T p.Q148* Chr.9: 21304815-21304815 on Build GRCh38
3656578 c.539A>G p.N180S Chr.9: 21304718-21304718 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368021464 Chr.9: 21304677-21304677 on Build GRCh38
rs931820018 Chr.9: 21304767-21304767 on Build GRCh38
rs10757212 Chr.9: 21304804-21304804 on Build GRCh38
rs778406590 Chr.9: 21304647-21304647 on Build GRCh38
rs757707162 Chr.9: 21304768-21304768 on Build GRCh38
rs146740793 Chr.9: 21304735-21304735 on Build GRCh38
rs780031657 Chr.9: 21304671-21304671 on Build GRCh38
rs373970079 Chr.9: 21304646-21304646 on Build GRCh38
rs779644580 Chr.9: 21304760-21304760 on Build GRCh38
rs759938906 Chr.9: 21304709-21304709 on Build GRCh38
rs150951978 Chr.9: 21304650-21304650 on Build GRCh38
rs763411775 Chr.9: 21304732-21304732 on Build GRCh38
rs377546833 Chr.9: 21304733-21304733 on Build GRCh38
rs769690428 Chr.9: 21304670-21304670 on Build GRCh38
rs367774355 Chr.9: 21304791-21304791 on Build GRCh38
rs747629817 Chr.9: 21304776-21304776 on Build GRCh38
rs755954027 Chr.9: 21304752-21304752 on Build GRCh38
rs370984273 Chr.9: 21304657-21304657 on Build GRCh38
rs557350282 Chr.9: 21304751-21304751 on Build GRCh38
rs764554251 Chr.9: 21304795-21304795 on Build GRCh38
rs781014511 Chr.9: 21304658-21304658 on Build GRCh38
rs763501265 Chr.9: 21304793-21304793 on Build GRCh38
rs772904137 Chr.9: 21304787-21304787 on Build GRCh38
rs1054114336 Chr.9: 21304759-21304759 on Build GRCh38
rs767440710 Chr.9: 21304803-21304803 on Build GRCh38
rs765742464 Chr.9: 21304649-21304649 on Build GRCh38
rs778611433 Chr.9: 21304773-21304773 on Build GRCh38
rs970344223 Chr.9: 21304700-21304700 on Build GRCh38
rs760009555 Chr.9: 21304648-21304648 on Build GRCh38
rs150500794 Chr.9: 21304689-21304689 on Build GRCh38
rs758457502 Chr.9: 21304652-21304652 on Build GRCh38
rs374778354 Chr.9: 21304783-21304783 on Build GRCh38
rs756247344 Chr.9: 21304811-21304811 on Build GRCh38
rs777766332 Chr.9: 21304654-21304654 on Build GRCh38
rs920400338 Chr.9: 21304696-21304696 on Build GRCh38
rs140942044 Chr.9: 21304681-21304681 on Build GRCh38
rs745649623 Chr.9: 21304659-21304659 on Build GRCh38
rs41298200 Chr.9: 21304797-21304797 on Build GRCh38
rs761871945 Chr.9: 21304801-21304801 on Build GRCh38
rs144769373 Chr.9: 21304721-21304721 on Build GRCh38
rs751813146 Chr.9: 21304655-21304655 on Build GRCh38
rs768284296 Chr.9: 21304680-21304680 on Build GRCh38
rs921753010 Chr.9: 21304762-21304762 on Build GRCh38
rs370774278 Chr.9: 21304737-21304737 on Build GRCh38
rs746582961 Chr.9: 21304789-21304789 on Build GRCh38
rs375863195 Chr.9: 21304684-21304684 on Build GRCh38

View Full Product Details