Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 134493405-134493664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPRT1
Entrez Gene ID
3251
Gene Name
hypoxanthine phosphoribosyltransferase 1
Gene Aliases
HGPRT, HPRT
UniGene
Hs.412707
Gene Chromosome Location
Chr.X: 134460145-134500668 on Build GRCh38
Transcripts
NM_000194.2

SNP Detail

SNP ID Chromosome Location
rs374407642 Chr.X: 134493481-134493481 on Build GRCh38
rs137852505 Chr.X: 134493564-134493564 on Build GRCh38
rs745871049 Chr.X: 134493561-134493561 on Build GRCh38
rs750725490 Chr.X: 134493608-134493609 on Build GRCh38
rs137852484 Chr.X: 134493586-134493586 on Build GRCh38
rs886042455 Chr.X: 134493590-134493590 on Build GRCh38
rs752496213 Chr.X: 134493503-134493503 on Build GRCh38
rs767466109 Chr.X: 134493440-134493440 on Build GRCh38
rs764842798 Chr.X: 134493461-134493461 on Build GRCh38
rs137852496 Chr.X: 134493533-134493533 on Build GRCh38
rs905921577 Chr.X: 134493454-134493454 on Build GRCh38
rs746094095 Chr.X: 134493546-134493546 on Build GRCh38
rs137852503 Chr.X: 134493524-134493524 on Build GRCh38
rs757630214 Chr.X: 134493525-134493525 on Build GRCh38
rs777634183 Chr.X: 134493627-134493627 on Build GRCh38
rs148780933 Chr.X: 134493585-134493585 on Build GRCh38
rs1020538716 Chr.X: 134493593-134493593 on Build GRCh38
rs749320387 Chr.X: 134493615-134493615 on Build GRCh38
rs371416162 Chr.X: 134493458-134493458 on Build GRCh38
rs781608428 Chr.X: 134493537-134493537 on Build GRCh38
rs752527618 Chr.X: 134493588-134493588 on Build GRCh38
rs756264380 Chr.X: 134493549-134493549 on Build GRCh38
rs370309875 Chr.X: 134493619-134493619 on Build GRCh38
rs376266868 Chr.X: 134493491-134493491 on Build GRCh38
rs1000513891 Chr.X: 134493632-134493632 on Build GRCh38
rs781248524 Chr.X: 134493443-134493443 on Build GRCh38
rs201976755 Chr.X: 134493496-134493496 on Build GRCh38
rs752213035 Chr.X: 134493474-134493474 on Build GRCh38

View Full Product Details