Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 46895734-46895959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTR2A
Entrez Gene ID
3356
Gene Name
5-hydroxytryptamine receptor 2A
Gene Aliases
5-HT2A, HTR2
UniGene
Hs.72630
Gene Chromosome Location
Chr.13: 46831542-46897076 on Build GRCh38
Transcripts
NM_000621.4 NM_001165947.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5645148 c.26C>G p.T9S Chr.13: 46895881-46895881 on Build GRCh38
261343 c.29C>A p.S10Y Chr.13: 46895878-46895878 on Build GRCh38
1706768 c.155G>A p.R52Q Chr.13: 46895752-46895752 on Build GRCh38
5842601 c.31T>A p.L11M Chr.13: 46895876-46895876 on Build GRCh38
947760 c.136A>G p.T46A Chr.13: 46895771-46895771 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749889591 Chr.13: 46895760-46895760 on Build GRCh38
rs545813583 Chr.13: 46895872-46895872 on Build GRCh38
rs375024989 Chr.13: 46895793-46895793 on Build GRCh38
rs1805055 Chr.13: 46895833-46895833 on Build GRCh38
rs750120780 Chr.13: 46895850-46895850 on Build GRCh38
rs781107449 Chr.13: 46895804-46895805 on Build GRCh38
rs755545262 Chr.13: 46895793-46895807 on Build GRCh38
rs747855847 Chr.13: 46895867-46895867 on Build GRCh38
rs778501828 Chr.13: 46895789-46895789 on Build GRCh38
rs762264146 Chr.13: 46895839-46895839 on Build GRCh38
rs746149691 Chr.13: 46895898-46895898 on Build GRCh38
rs150684157 Chr.13: 46895856-46895856 on Build GRCh38
rs139775104 Chr.13: 46895826-46895826 on Build GRCh38
rs1046592807 Chr.13: 46895804-46895804 on Build GRCh38
rs532924268 Chr.13: 46895900-46895900 on Build GRCh38
rs777023991 Chr.13: 46895816-46895816 on Build GRCh38
rs770043192 Chr.13: 46895822-46895822 on Build GRCh38
rs201794484 Chr.13: 46895766-46895766 on Build GRCh38
rs781542336 Chr.13: 46895877-46895877 on Build GRCh38
rs1052540084 Chr.13: 46895851-46895851 on Build GRCh38
rs979444009 Chr.13: 46895914-46895914 on Build GRCh38
rs765936117 Chr.13: 46895858-46895858 on Build GRCh38
rs748483027 Chr.13: 46895891-46895891 on Build GRCh38
rs778929461 Chr.13: 46895866-46895866 on Build GRCh38
rs769990169 Chr.13: 46895915-46895915 on Build GRCh38
rs117554656 Chr.13: 46895924-46895924 on Build GRCh38
rs755654778 Chr.13: 46895765-46895765 on Build GRCh38
rs768489549 Chr.13: 46895788-46895788 on Build GRCh38
rs185040200 Chr.13: 46895846-46895846 on Build GRCh38
rs745909918 Chr.13: 46895819-46895819 on Build GRCh38
rs749275506 Chr.13: 46895770-46895770 on Build GRCh38
rs754812239 Chr.13: 46895863-46895863 on Build GRCh38
rs1045024958 Chr.13: 46895792-46895792 on Build GRCh38
rs201460128 Chr.13: 46895923-46895923 on Build GRCh38
rs868475357 Chr.13: 46895801-46895801 on Build GRCh38
rs577997891 Chr.13: 46895862-46895862 on Build GRCh38
rs6313 Chr.13: 46895805-46895805 on Build GRCh38
rs774523543 Chr.13: 46895837-46895837 on Build GRCh38

View Full Product Details