Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119047529-119047790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HYOU1
Entrez Gene ID
10525
Gene Name
hypoxia up-regulated 1
Gene Aliases
GRP-170, Grp170, HSP12A, ORP-150, ORP150
UniGene
Hs.277704
Gene Chromosome Location
Chr.11: 119044185-119057246 on Build GRCh38
Transcripts
NM_001130991.2 NM_006389.4 XM_005271392.3 XM_005271393.3 XM_005271394.3 XM_017017095.1 XM_017017096.1 XM_017017097.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4760599 c.2572G>C p.E858Q Chr.11: 119047757-119047757 on Build GRCh38
5899620 c.2557G>T p.E853* Chr.11: 119047772-119047772 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1008748428 Chr.11: 119047664-119047664 on Build GRCh38
rs201897343 Chr.11: 119047701-119047701 on Build GRCh38
rs918500768 Chr.11: 119047602-119047602 on Build GRCh38
rs377610698 Chr.11: 119047705-119047705 on Build GRCh38
rs782714818 Chr.11: 119047703-119047704 on Build GRCh38
rs34470243 Chr.11: 119047708-119047709 on Build GRCh38
rs371646359 Chr.11: 119047754-119047754 on Build GRCh38
rs782257624 Chr.11: 119047762-119047762 on Build GRCh38
rs781908039 Chr.11: 119047710-119047710 on Build GRCh38
rs536387318 Chr.11: 119047682-119047682 on Build GRCh38
rs782198949 Chr.11: 119047726-119047726 on Build GRCh38
rs782402499 Chr.11: 119047696-119047696 on Build GRCh38
rs140462408 Chr.11: 119047690-119047690 on Build GRCh38
rs1019672576 Chr.11: 119047668-119047668 on Build GRCh38
rs924787266 Chr.11: 119047727-119047727 on Build GRCh38
rs550726137 Chr.11: 119047637-119047637 on Build GRCh38
rs899083333 Chr.11: 119047558-119047558 on Build GRCh38
rs150386978 Chr.11: 119047629-119047629 on Build GRCh38
rs912190885 Chr.11: 119047761-119047761 on Build GRCh38
rs782538718 Chr.11: 119047715-119047715 on Build GRCh38
rs752015250 Chr.11: 119047635-119047635 on Build GRCh38
rs138137230 Chr.11: 119047638-119047638 on Build GRCh38
rs1037615513 Chr.11: 119047554-119047554 on Build GRCh38
rs151204513 Chr.11: 119047559-119047559 on Build GRCh38
rs782654818 Chr.11: 119047722-119047722 on Build GRCh38
rs546496561 Chr.11: 119047566-119047566 on Build GRCh38
rs782370481 Chr.11: 119047765-119047765 on Build GRCh38
rs969358280 Chr.11: 119047703-119047703 on Build GRCh38
rs781822973 Chr.11: 119047762-119047764 on Build GRCh38
rs782106819 Chr.11: 119047702-119047702 on Build GRCh38
rs140378199 Chr.11: 119047607-119047607 on Build GRCh38
rs782699897 Chr.11: 119047709-119047709 on Build GRCh38
rs782105820 Chr.11: 119047761-119047762 on Build GRCh38
rs782780774 Chr.11: 119047766-119047766 on Build GRCh38
rs782757061 Chr.11: 119047704-119047704 on Build GRCh38
rs782484662 Chr.11: 119047723-119047723 on Build GRCh38
rs534349629 Chr.11: 119047744-119047744 on Build GRCh38

View Full Product Details