Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130075539-130075788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR107
Entrez Gene ID
57720
Gene Name
G protein-coupled receptor 107
Gene Aliases
GCDRP, LUSTR1, bA138E2.2
UniGene
Hs.512461
Gene Chromosome Location
Chr.9: 130053706-130140169 on Build GRCh38
Transcripts
NM_001136557.1 NM_001136558.1 NM_001287346.1 NM_020960.4 XM_017014953.1 XM_017014954.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7284615 c.233A>G p.E78G Chr.9: 130075727-130075727 on Build GRCh38
6673406 c.255+1G>A p.? Chr.9: 130075750-130075750 on Build GRCh38
4407431 c.243C>T p.D81D Chr.9: 130075737-130075737 on Build GRCh38
6182793 c.188A>T p.K63M Chr.9: 130075682-130075682 on Build GRCh38
6182794 c.188A>T p.K63M Chr.9: 130075682-130075682 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368673169 Chr.9: 130075746-130075746 on Build GRCh38
rs758782922 Chr.9: 130075622-130075622 on Build GRCh38
rs537886231 Chr.9: 130075633-130075633 on Build GRCh38
rs528452332 Chr.9: 130075650-130075650 on Build GRCh38
rs759619148 Chr.9: 130075739-130075739 on Build GRCh38
rs557763083 Chr.9: 130075642-130075642 on Build GRCh38
rs747417381 Chr.9: 130075628-130075628 on Build GRCh38
rs757760401 Chr.9: 130075757-130075757 on Build GRCh38
rs766640091 Chr.9: 130075601-130075601 on Build GRCh38
rs371130458 Chr.9: 130075748-130075748 on Build GRCh38
rs773674300 Chr.9: 130075670-130075670 on Build GRCh38
rs577845310 Chr.9: 130075680-130075680 on Build GRCh38
rs755119568 Chr.9: 130075610-130075610 on Build GRCh38
rs752679373 Chr.9: 130075743-130075743 on Build GRCh38
rs773115029 Chr.9: 130075689-130075689 on Build GRCh38
rs752098424 Chr.9: 130075756-130075756 on Build GRCh38
rs141390611 Chr.9: 130075720-130075720 on Build GRCh38
rs760435037 Chr.9: 130075597-130075597 on Build GRCh38
rs753262523 Chr.9: 130075615-130075615 on Build GRCh38
rs569162175 Chr.9: 130075577-130075577 on Build GRCh38
rs768213512 Chr.9: 130075660-130075660 on Build GRCh38
rs761584859 Chr.9: 130075587-130075587 on Build GRCh38
rs949642056 Chr.9: 130075704-130075704 on Build GRCh38
rs754048933 Chr.9: 130075602-130075602 on Build GRCh38
rs766160806 Chr.9: 130075709-130075709 on Build GRCh38
rs374428340 Chr.9: 130075681-130075681 on Build GRCh38
rs748942096 Chr.9: 130075657-130075657 on Build GRCh38
rs765381982 Chr.9: 130075740-130075740 on Build GRCh38
rs376703202 Chr.9: 130075702-130075702 on Build GRCh38
rs779908757 Chr.9: 130075636-130075636 on Build GRCh38
rs750149103 Chr.9: 130075595-130075595 on Build GRCh38
rs73539696 Chr.9: 130075737-130075737 on Build GRCh38
rs778230660 Chr.9: 130075627-130075627 on Build GRCh38

View Full Product Details