Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 35867247-35867520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL7R
Entrez Gene ID
3575
Gene Name
interleukin 7 receptor
Gene Aliases
CD127, CDW127, IL-7R-alpha, IL7RA, ILRA
UniGene
Hs.591742
Gene Chromosome Location
Chr.5: 35856875-35879603 on Build GRCh38
Transcripts
NM_002185.3 NR_120485.1 XM_005248299.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6171023 c.377T>C p.I126T Chr.5: 35867461-35867461 on Build GRCh38
6920161 c.262C>A p.L88I Chr.5: 35867346-35867346 on Build GRCh38
1437173 c.337G>A p.E113K Chr.5: 35867421-35867421 on Build GRCh38
5403377 c.303G>A p.L101L Chr.5: 35867387-35867387 on Build GRCh38
1437172 c.270G>T p.E90D Chr.5: 35867354-35867354 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757797163 Chr.5: 35867329-35867329 on Build GRCh38
rs11567735 Chr.5: 35867423-35867423 on Build GRCh38
rs776441054 Chr.5: 35867392-35867392 on Build GRCh38
rs111370546 Chr.5: 35867471-35867471 on Build GRCh38
rs201058754 Chr.5: 35867399-35867399 on Build GRCh38
rs755030275 Chr.5: 35867311-35867311 on Build GRCh38
rs200506624 Chr.5: 35867442-35867442 on Build GRCh38
rs199641706 Chr.5: 35867417-35867417 on Build GRCh38
rs141698985 Chr.5: 35867349-35867349 on Build GRCh38
rs202194801 Chr.5: 35867284-35867284 on Build GRCh38
rs757709286 Chr.5: 35867409-35867409 on Build GRCh38
rs1020619032 Chr.5: 35867341-35867341 on Build GRCh38
rs759585748 Chr.5: 35867393-35867393 on Build GRCh38
rs552630274 Chr.5: 35867321-35867321 on Build GRCh38
rs778262602 Chr.5: 35867419-35867419 on Build GRCh38
rs754784201 Chr.5: 35867428-35867429 on Build GRCh38
rs140473900 Chr.5: 35867450-35867450 on Build GRCh38
rs548638151 Chr.5: 35867416-35867416 on Build GRCh38
rs200817423 Chr.5: 35867420-35867420 on Build GRCh38
rs894868423 Chr.5: 35867430-35867430 on Build GRCh38
rs762188432 Chr.5: 35867462-35867462 on Build GRCh38
rs771757728 Chr.5: 35867384-35867384 on Build GRCh38
rs200687605 Chr.5: 35867316-35867316 on Build GRCh38
rs771667604 Chr.5: 35867424-35867424 on Build GRCh38
rs760580267 Chr.5: 35867282-35867282 on Build GRCh38
rs537367054 Chr.5: 35867428-35867428 on Build GRCh38
rs191585195 Chr.5: 35867367-35867367 on Build GRCh38
rs193922641 Chr.5: 35867437-35867437 on Build GRCh38
rs772953676 Chr.5: 35867385-35867385 on Build GRCh38
rs869312857 Chr.5: 35867445-35867446 on Build GRCh38
rs919258603 Chr.5: 35867477-35867477 on Build GRCh38
rs150051812 Chr.5: 35867398-35867398 on Build GRCh38
rs201767916 Chr.5: 35867330-35867330 on Build GRCh38
rs753909264 Chr.5: 35867297-35867297 on Build GRCh38
rs777419876 Chr.5: 35867426-35867426 on Build GRCh38
rs751425932 Chr.5: 35867383-35867385 on Build GRCh38
rs751922549 Chr.5: 35867407-35867407 on Build GRCh38
rs1031921568 Chr.5: 35867467-35867467 on Build GRCh38
rs926877898 Chr.5: 35867497-35867497 on Build GRCh38
rs773973629 Chr.5: 35867374-35867374 on Build GRCh38
rs199742437 Chr.5: 35867397-35867397 on Build GRCh38
rs910624460 Chr.5: 35867277-35867277 on Build GRCh38
rs764440449 Chr.5: 35867403-35867403 on Build GRCh38
rs200475138 Chr.5: 35867323-35867323 on Build GRCh38
rs201106853 Chr.5: 35867391-35867391 on Build GRCh38
rs750846051 Chr.5: 35867473-35867473 on Build GRCh38
rs145407742 Chr.5: 35867457-35867457 on Build GRCh38
rs746743911 Chr.5: 35867427-35867427 on Build GRCh38
rs193922640 Chr.5: 35867364-35867365 on Build GRCh38
rs781675111 Chr.5: 35867410-35867410 on Build GRCh38
rs971681884 Chr.5: 35867458-35867458 on Build GRCh38
rs377557867 Chr.5: 35867488-35867488 on Build GRCh38
rs773047299 Chr.5: 35867271-35867271 on Build GRCh38
rs769810965 Chr.5: 35867447-35867447 on Build GRCh38
rs929178646 Chr.5: 35867380-35867380 on Build GRCh38
rs758304802 Chr.5: 35867283-35867284 on Build GRCh38
rs201710684 Chr.5: 35867443-35867443 on Build GRCh38
rs201643973 Chr.5: 35867487-35867487 on Build GRCh38
rs201207454 Chr.5: 35867460-35867460 on Build GRCh38
rs139835773 Chr.5: 35867286-35867286 on Build GRCh38
rs200241281 Chr.5: 35867390-35867390 on Build GRCh38
rs778067695 Chr.5: 35867366-35867366 on Build GRCh38
rs766399441 Chr.5: 35867314-35867314 on Build GRCh38
rs1049267335 Chr.5: 35867496-35867496 on Build GRCh38
rs756545602 Chr.5: 35867479-35867479 on Build GRCh38
rs747417170 Chr.5: 35867327-35867327 on Build GRCh38

View Full Product Details