Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15117838-15118075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ILVBL
Entrez Gene ID
10994
Gene Name
ilvB acetolactate synthase like
Gene Aliases
209L8, AHAS, HACL1L, ILV2H
UniGene
Hs.78880
Gene Chromosome Location
Chr.19: 15114974-15125799 on Build GRCh38
Transcripts
NM_006844.4 XM_005259717.4 XM_011527651.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3970759 c.1063G>C p.D355H Chr.19: 15118004-15118004 on Build GRCh38
6309538 c.1124G>A p.R375H Chr.19: 15117943-15117943 on Build GRCh38
992043 c.1070G>A p.R357H Chr.19: 15117997-15117997 on Build GRCh38
5854843 c.1164G>A p.W388* Chr.19: 15117903-15117903 on Build GRCh38
7114385 c.1113C>G p.I371M Chr.19: 15117954-15117954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs564387950 Chr.19: 15117861-15117861 on Build GRCh38
rs373266564 Chr.19: 15117860-15117860 on Build GRCh38
rs770984066 Chr.19: 15117998-15117998 on Build GRCh38
rs918303619 Chr.19: 15117866-15117866 on Build GRCh38
rs768580763 Chr.19: 15117960-15117960 on Build GRCh38
rs760827838 Chr.19: 15117997-15117997 on Build GRCh38
rs765821594 Chr.19: 15118019-15118019 on Build GRCh38
rs115688072 Chr.19: 15117946-15117946 on Build GRCh38
rs370515393 Chr.19: 15117876-15117876 on Build GRCh38
rs200540246 Chr.19: 15117875-15117875 on Build GRCh38
rs760779811 Chr.19: 15117895-15117895 on Build GRCh38
rs796533630 Chr.19: 15117928-15117928 on Build GRCh38
rs755604021 Chr.19: 15117951-15117951 on Build GRCh38
rs142150131 Chr.19: 15117938-15117938 on Build GRCh38
rs896087736 Chr.19: 15117950-15117950 on Build GRCh38
rs968222194 Chr.19: 15117995-15117995 on Build GRCh38
rs779556139 Chr.19: 15117953-15117953 on Build GRCh38
rs779752877 Chr.19: 15118040-15118040 on Build GRCh38
rs763343004 Chr.19: 15118025-15118025 on Build GRCh38
rs764394425 Chr.19: 15118027-15118027 on Build GRCh38
rs536567026 Chr.19: 15117944-15117944 on Build GRCh38
rs537228490 Chr.19: 15118023-15118023 on Build GRCh38
rs747953395 Chr.19: 15117966-15117966 on Build GRCh38
rs777535239 Chr.19: 15117951-15117953 on Build GRCh38
rs755729406 Chr.19: 15118039-15118039 on Build GRCh38
rs775936422 Chr.19: 15118022-15118023 on Build GRCh38
rs761663550 Chr.19: 15117882-15117882 on Build GRCh38
rs377175719 Chr.19: 15117868-15117868 on Build GRCh38
rs931060392 Chr.19: 15117872-15117872 on Build GRCh38
rs559057427 Chr.19: 15118042-15118042 on Build GRCh38
rs17856373 Chr.19: 15117947-15117947 on Build GRCh38
rs367743686 Chr.19: 15118037-15118037 on Build GRCh38
rs762978112 Chr.19: 15117891-15117891 on Build GRCh38
rs749248897 Chr.19: 15117864-15117864 on Build GRCh38
rs1008914139 Chr.19: 15118041-15118041 on Build GRCh38
rs977924248 Chr.19: 15118009-15118009 on Build GRCh38
rs374699967 Chr.19: 15117893-15117893 on Build GRCh38
rs771803441 Chr.19: 15117983-15117983 on Build GRCh38
rs780872895 Chr.19: 15117945-15117945 on Build GRCh38
rs371179132 Chr.19: 15118049-15118049 on Build GRCh38
rs766398896 Chr.19: 15117920-15117920 on Build GRCh38
rs763727051 Chr.19: 15117907-15117907 on Build GRCh38
rs149772431 Chr.19: 15118011-15118011 on Build GRCh38
rs751413681 Chr.19: 15117937-15117937 on Build GRCh38
rs140967399 Chr.19: 15117959-15117959 on Build GRCh38
rs980137314 Chr.19: 15118044-15118045 on Build GRCh38
rs759756009 Chr.19: 15118018-15118018 on Build GRCh38
rs1022078552 Chr.19: 15117980-15117980 on Build GRCh38
rs576266378 Chr.19: 15117996-15117996 on Build GRCh38

View Full Product Details