Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 156496109-156496354 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPD2
Entrez Gene ID
2820
Gene Name
glycerol-3-phosphate dehydrogenase 2
Gene Aliases
GDH2, GPDM, mGPDH
UniGene
Hs.512382
Gene Chromosome Location
Chr.2: 156435290-156586403 on Build GRCh38
Transcripts
NM_000408.4 NM_001083112.2 XM_005246469.2 XM_011510977.2 XM_011510978.2 XM_017003830.1 XM_017003831.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7154815 c.203C>G p.S68C Chr.2: 156496144-156496144 on Build GRCh38
6364557 c.191T>C p.L64S Chr.2: 156496132-156496132 on Build GRCh38
1007984 c.229G>T p.G77* Chr.2: 156496170-156496170 on Build GRCh38
1613538 c.274+8delT p.? Chr.2: 156496223-156496223 on Build GRCh38
4409029 c.218T>A p.L73H Chr.2: 156496159-156496159 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs926577068 Chr.2: 156496322-156496322 on Build GRCh38
rs923417435 Chr.2: 156496209-156496209 on Build GRCh38
rs746961151 Chr.2: 156496293-156496293 on Build GRCh38
rs188328187 Chr.2: 156496234-156496234 on Build GRCh38
rs754127044 Chr.2: 156496235-156496235 on Build GRCh38
rs772975270 Chr.2: 156496202-156496202 on Build GRCh38
rs546273294 Chr.2: 156496317-156496317 on Build GRCh38
rs932085569 Chr.2: 156496229-156496229 on Build GRCh38
rs758346865 Chr.2: 156496216-156496217 on Build GRCh38
rs55843224 Chr.2: 156496234-156496234 on Build GRCh38
rs75842291 Chr.2: 156496224-156496224 on Build GRCh38
rs768082223 Chr.2: 156496230-156496230 on Build GRCh38
rs575600672 Chr.2: 156496259-156496259 on Build GRCh38
rs778983823 Chr.2: 156496261-156496261 on Build GRCh38
rs145159302 Chr.2: 156496136-156496136 on Build GRCh38
rs200015844 Chr.2: 156496232-156496232 on Build GRCh38
rs77492341 Chr.2: 156496142-156496142 on Build GRCh38
rs903068842 Chr.2: 156496271-156496271 on Build GRCh38
rs771088432 Chr.2: 156496172-156496172 on Build GRCh38
rs753118584 Chr.2: 156496231-156496231 on Build GRCh38
rs774380282 Chr.2: 156496181-156496181 on Build GRCh38
rs751076393 Chr.2: 156496206-156496206 on Build GRCh38
rs750454832 Chr.2: 156496195-156496196 on Build GRCh38
rs777258380 Chr.2: 156496235-156496235 on Build GRCh38
rs768527779 Chr.2: 156496183-156496183 on Build GRCh38
rs1002679857 Chr.2: 156496277-156496277 on Build GRCh38
rs762492206 Chr.2: 156496204-156496204 on Build GRCh38
rs777604222 Chr.2: 156496239-156496239 on Build GRCh38
rs747813747 Chr.2: 156496230-156496235 on Build GRCh38
rs776422379 Chr.2: 156496190-156496190 on Build GRCh38
rs766291257 Chr.2: 156496223-156496223 on Build GRCh38
rs751264242 Chr.2: 156496222-156496223 on Build GRCh38
rs760143793 Chr.2: 156496223-156496223 on Build GRCh38
rs988214746 Chr.2: 156496260-156496260 on Build GRCh38
rs957036499 Chr.2: 156496235-156496240 on Build GRCh38
rs748476035 Chr.2: 156496163-156496163 on Build GRCh38
rs749469985 Chr.2: 156496165-156496165 on Build GRCh38
rs200997719 Chr.2: 156496214-156496214 on Build GRCh38
rs754690876 Chr.2: 156496223-156496224 on Build GRCh38
rs115026480 Chr.2: 156496203-156496203 on Build GRCh38
rs761600581 Chr.2: 156496195-156496195 on Build GRCh38
rs145747966 Chr.2: 156496164-156496164 on Build GRCh38
rs567802812 Chr.2: 156496231-156496235 on Build GRCh38
rs746976112 Chr.2: 156496182-156496182 on Build GRCh38
rs894298821 Chr.2: 156496327-156496327 on Build GRCh38
rs764937237 Chr.2: 156496196-156496196 on Build GRCh38

View Full Product Details