Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 116683642-116683867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA12A
Entrez Gene ID
259217
Gene Name
heat shock protein family A (Hsp70) member 12A
Gene Aliases
-
UniGene
Hs.648448
Gene Chromosome Location
Chr.10: 116671192-116851241 on Build GRCh38
Transcripts
NM_025015.2 XM_005269672.3 XM_005269673.4 XM_011539579.2 XM_011539580.2 XM_017016032.1

SNP Detail

SNP ID Chromosome Location
rs3740575 Chr.10: 116683752-116683752 on Build GRCh38
rs782225880 Chr.10: 116683708-116683708 on Build GRCh38
rs781989904 Chr.10: 116683728-116683728 on Build GRCh38
rs781963076 Chr.10: 116683749-116683749 on Build GRCh38
rs782420092 Chr.10: 116683828-116683828 on Build GRCh38
rs782689381 Chr.10: 116683787-116683787 on Build GRCh38
rs559548706 Chr.10: 116683819-116683819 on Build GRCh38
rs563679259 Chr.10: 116683709-116683709 on Build GRCh38
rs782039705 Chr.10: 116683751-116683751 on Build GRCh38
rs782425211 Chr.10: 116683727-116683727 on Build GRCh38
rs202216924 Chr.10: 116683831-116683831 on Build GRCh38
rs782064654 Chr.10: 116683839-116683839 on Build GRCh38
rs181157543 Chr.10: 116683674-116683674 on Build GRCh38
rs782729536 Chr.10: 116683747-116683747 on Build GRCh38
rs782216597 Chr.10: 116683810-116683810 on Build GRCh38
rs782659363 Chr.10: 116683685-116683685 on Build GRCh38
rs41284380 Chr.10: 116683801-116683801 on Build GRCh38
rs782164334 Chr.10: 116683759-116683759 on Build GRCh38
rs782760552 Chr.10: 116683765-116683765 on Build GRCh38
rs782561774 Chr.10: 116683774-116683774 on Build GRCh38
rs953321804 Chr.10: 116683746-116683746 on Build GRCh38
rs781806869 Chr.10: 116683778-116683778 on Build GRCh38
rs372981337 Chr.10: 116683830-116683830 on Build GRCh38
rs781952236 Chr.10: 116683770-116683771 on Build GRCh38
rs782494872 Chr.10: 116683683-116683683 on Build GRCh38
rs370309428 Chr.10: 116683832-116683832 on Build GRCh38
rs782810578 Chr.10: 116683840-116683840 on Build GRCh38
rs782609994 Chr.10: 116683808-116683808 on Build GRCh38
rs553554607 Chr.10: 116683784-116683784 on Build GRCh38
rs781929997 Chr.10: 116683824-116683824 on Build GRCh38
rs781937678 Chr.10: 116683713-116683713 on Build GRCh38
rs782295535 Chr.10: 116683677-116683677 on Build GRCh38
rs782139430 Chr.10: 116683825-116683825 on Build GRCh38
rs781791082 Chr.10: 116683836-116683836 on Build GRCh38
rs560978815 Chr.10: 116683772-116683773 on Build GRCh38
rs782135214 Chr.10: 116683737-116683737 on Build GRCh38
rs782531675 Chr.10: 116683786-116683786 on Build GRCh38
rs782359360 Chr.10: 116683685-116683685 on Build GRCh38
rs376697507 Chr.10: 116683720-116683720 on Build GRCh38
rs781861635 Chr.10: 116683673-116683673 on Build GRCh38
rs376261288 Chr.10: 116683793-116683793 on Build GRCh38
rs200396979 Chr.10: 116683804-116683804 on Build GRCh38

View Full Product Details