Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 19861744-19862007 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPRC5B
Entrez Gene ID
51704
Gene Name
G protein-coupled receptor class C group 5 member B
Gene Aliases
RAIG-2, RAIG2
UniGene
Hs.148685
Gene Chromosome Location
Chr.16: 19858971-19886045 on Build GRCh38
Transcripts
NM_001304771.1 NM_016235.2 XM_006721051.2 XM_006721052.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5519369 c.1031-3delC p.? Chr.16: 19861976-19861976 on Build GRCh38
6268759 c.1158C>A p.N386K Chr.16: 19861846-19861846 on Build GRCh38
6555472 c.1104C>A p.S368R Chr.16: 19861900-19861900 on Build GRCh38
7303164 c.1097G>T p.R366I Chr.16: 19861907-19861907 on Build GRCh38
1208785 c.1099C>T p.P367S Chr.16: 19861905-19861905 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748535417 Chr.16: 19861867-19861867 on Build GRCh38
rs778431012 Chr.16: 19861939-19861939 on Build GRCh38
rs778065622 Chr.16: 19861900-19861901 on Build GRCh38
rs776604761 Chr.16: 19861901-19861901 on Build GRCh38
rs533784987 Chr.16: 19861821-19861821 on Build GRCh38
rs771612264 Chr.16: 19861978-19861981 on Build GRCh38
rs766687109 Chr.16: 19861764-19861764 on Build GRCh38
rs377092288 Chr.16: 19861896-19861896 on Build GRCh38
rs774745807 Chr.16: 19861986-19861986 on Build GRCh38
rs759744621 Chr.16: 19861918-19861918 on Build GRCh38
rs2074427 Chr.16: 19861785-19861785 on Build GRCh38
rs369976914 Chr.16: 19861828-19861828 on Build GRCh38
rs771728943 Chr.16: 19861895-19861895 on Build GRCh38
rs760463381 Chr.16: 19861899-19861899 on Build GRCh38
rs140153375 Chr.16: 19861968-19861968 on Build GRCh38
rs371638316 Chr.16: 19861973-19861973 on Build GRCh38
rs546183420 Chr.16: 19861927-19861927 on Build GRCh38
rs778323081 Chr.16: 19861928-19861928 on Build GRCh38
rs770445030 Chr.16: 19861790-19861790 on Build GRCh38
rs61746631 Chr.16: 19861855-19861855 on Build GRCh38
rs199692459 Chr.16: 19861900-19861900 on Build GRCh38
rs201199981 Chr.16: 19861795-19861795 on Build GRCh38
rs562433136 Chr.16: 19861789-19861789 on Build GRCh38
rs148962392 Chr.16: 19861858-19861858 on Build GRCh38
rs751611822 Chr.16: 19861917-19861917 on Build GRCh38
rs567291041 Chr.16: 19861913-19861936 on Build GRCh38
rs771641063 Chr.16: 19861958-19861958 on Build GRCh38
rs149762584 Chr.16: 19861846-19861846 on Build GRCh38
rs761693107 Chr.16: 19861904-19861904 on Build GRCh38
rs757728423 Chr.16: 19861936-19861936 on Build GRCh38
rs78603358 Chr.16: 19861778-19861778 on Build GRCh38
rs1033579139 Chr.16: 19861977-19861977 on Build GRCh38
rs751771117 Chr.16: 19861857-19861857 on Build GRCh38
rs778420947 Chr.16: 19861881-19861881 on Build GRCh38
rs146737474 Chr.16: 19861948-19861948 on Build GRCh38
rs993401527 Chr.16: 19861781-19861781 on Build GRCh38
rs946370746 Chr.16: 19861859-19861859 on Build GRCh38
rs1026758104 Chr.16: 19861783-19861783 on Build GRCh38
rs754220843 Chr.16: 19861777-19861777 on Build GRCh38
rs147960290 Chr.16: 19861894-19861894 on Build GRCh38
rs770444643 Chr.16: 19861889-19861889 on Build GRCh38
rs746717252 Chr.16: 19861967-19861967 on Build GRCh38
rs375374151 Chr.16: 19861924-19861924 on Build GRCh38
rs749809226 Chr.16: 19861882-19861882 on Build GRCh38
rs756622372 Chr.16: 19861879-19861879 on Build GRCh38
rs201722584 Chr.16: 19861912-19861912 on Build GRCh38
rs550314452 Chr.16: 19861786-19861786 on Build GRCh38
rs549155662 Chr.16: 19861854-19861854 on Build GRCh38
rs373407280 Chr.16: 19861845-19861845 on Build GRCh38
rs775287947 Chr.16: 19861837-19861837 on Build GRCh38
rs887783254 Chr.16: 19861784-19861784 on Build GRCh38
rs533922181 Chr.16: 19861926-19861926 on Build GRCh38
rs753034705 Chr.16: 19861925-19861925 on Build GRCh38
rs773766558 Chr.16: 19861890-19861890 on Build GRCh38
rs200239467 Chr.16: 19861791-19861791 on Build GRCh38
rs111876397 Chr.16: 19861963-19861963 on Build GRCh38
rs754331652 Chr.16: 19861929-19861929 on Build GRCh38
rs113634112 Chr.16: 19861801-19861801 on Build GRCh38

View Full Product Details