Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26045516-26045773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIST1H3C
Entrez Gene ID
8352
Gene Name
histone cluster 1, H3c
Gene Aliases
H3.1, H3/c, H3FC
UniGene
Hs.248176
Gene Chromosome Location
Chr.6: 26045411-26045869 on Build GRCh38
Transcripts
NM_003531.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4983164 c.348A>G p.K116K Chr.6: 26045758-26045758 on Build GRCh38
6934476 c.209G>A p.R70H Chr.6: 26045619-26045619 on Build GRCh38
4880055 c.110A>G p.K37R Chr.6: 26045520-26045520 on Build GRCh38
6929897 c.166C>T p.Q56* Chr.6: 26045576-26045576 on Build GRCh38
1076684 c.149G>A p.R50H Chr.6: 26045559-26045559 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561705295 Chr.6: 26045642-26045642 on Build GRCh38
rs757321049 Chr.6: 26045713-26045713 on Build GRCh38
rs766815015 Chr.6: 26045555-26045555 on Build GRCh38
rs761955716 Chr.6: 26045739-26045740 on Build GRCh38
rs749935174 Chr.6: 26045641-26045641 on Build GRCh38
rs373098650 Chr.6: 26045545-26045545 on Build GRCh38
rs1042668277 Chr.6: 26045538-26045538 on Build GRCh38
rs375603729 Chr.6: 26045662-26045662 on Build GRCh38
rs143026777 Chr.6: 26045626-26045626 on Build GRCh38
rs771631277 Chr.6: 26045674-26045674 on Build GRCh38
rs753651183 Chr.6: 26045656-26045656 on Build GRCh38
rs149070402 Chr.6: 26045624-26045624 on Build GRCh38
rs764962871 Chr.6: 26045695-26045695 on Build GRCh38
rs779162034 Chr.6: 26045611-26045611 on Build GRCh38
rs771371857 Chr.6: 26045621-26045621 on Build GRCh38
rs766237552 Chr.6: 26045644-26045644 on Build GRCh38
rs3752416 Chr.6: 26045701-26045701 on Build GRCh38
rs527289737 Chr.6: 26045666-26045666 on Build GRCh38
rs200068681 Chr.6: 26045594-26045594 on Build GRCh38
rs147001758 Chr.6: 26045680-26045680 on Build GRCh38
rs9461231 Chr.6: 26045711-26045711 on Build GRCh38
rs777916771 Chr.6: 26045708-26045708 on Build GRCh38
rs1001071216 Chr.6: 26045591-26045591 on Build GRCh38
rs756894393 Chr.6: 26045599-26045599 on Build GRCh38
rs144926546 Chr.6: 26045602-26045602 on Build GRCh38
rs202100651 Chr.6: 26045596-26045596 on Build GRCh38
rs558243677 Chr.6: 26045572-26045572 on Build GRCh38
rs755131918 Chr.6: 26045610-26045610 on Build GRCh38
rs370158525 Chr.6: 26045665-26045665 on Build GRCh38
rs781709095 Chr.6: 26045716-26045716 on Build GRCh38
rs747499521 Chr.6: 26045619-26045619 on Build GRCh38
rs752518862 Chr.6: 26045697-26045697 on Build GRCh38
rs779861931 Chr.6: 26045608-26045608 on Build GRCh38
rs754830352 Chr.6: 26045657-26045657 on Build GRCh38
rs767531748 Chr.6: 26045600-26045600 on Build GRCh38
rs917890383 Chr.6: 26045648-26045648 on Build GRCh38
rs1018182182 Chr.6: 26045615-26045615 on Build GRCh38
rs200228685 Chr.6: 26045747-26045747 on Build GRCh38
rs752964674 Chr.6: 26045663-26045663 on Build GRCh38
rs774188493 Chr.6: 26045630-26045630 on Build GRCh38
rs761388983 Chr.6: 26045633-26045633 on Build GRCh38
rs773570245 Chr.6: 26045616-26045616 on Build GRCh38
rs983273765 Chr.6: 26045635-26045635 on Build GRCh38
rs756032161 Chr.6: 26045606-26045606 on Build GRCh38
rs141335378 Chr.6: 26045548-26045548 on Build GRCh38
rs780501933 Chr.6: 26045733-26045733 on Build GRCh38
rs771458123 Chr.6: 26045670-26045670 on Build GRCh38
rs749479857 Chr.6: 26045542-26045542 on Build GRCh38
rs753809029 Chr.6: 26045687-26045687 on Build GRCh38
rs766332675 Chr.6: 26045686-26045686 on Build GRCh38
rs771352320 Chr.6: 26045673-26045673 on Build GRCh38
rs762653599 Chr.6: 26045587-26045587 on Build GRCh38
rs1045347939 Chr.6: 26045566-26045566 on Build GRCh38
rs759300732 Chr.6: 26045689-26045689 on Build GRCh38
rs749844493 Chr.6: 26045552-26045552 on Build GRCh38
rs905351188 Chr.6: 26045581-26045581 on Build GRCh38
rs201464639 Chr.6: 26045671-26045671 on Build GRCh38
rs145110843 Chr.6: 26045554-26045554 on Build GRCh38
rs955045739 Chr.6: 26045609-26045609 on Build GRCh38
rs3752417 Chr.6: 26045677-26045677 on Build GRCh38
rs747063074 Chr.6: 26045668-26045668 on Build GRCh38
rs1019927798 Chr.6: 26045605-26045605 on Build GRCh38
rs749687894 Chr.6: 26045743-26045743 on Build GRCh38
rs576312429 Chr.6: 26045614-26045614 on Build GRCh38
rs746331871 Chr.6: 26045722-26045722 on Build GRCh38
rs772803858 Chr.6: 26045749-26045749 on Build GRCh38
rs748237932 Chr.6: 26045551-26045551 on Build GRCh38
rs767008919 Chr.6: 26045638-26045638 on Build GRCh38

View Full Product Details