Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 81705207-81705466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HGF
Entrez Gene ID
3082
Gene Name
hepatocyte growth factor
Gene Aliases
DFNB39, F-TCF, HGFB, HPTA, SF
UniGene
Hs.396530
Gene Chromosome Location
Chr.7: 81699006-81770438 on Build GRCh38
Transcripts
NM_000601.5 NM_001010932.2 XM_006715956.2 XM_011516115.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3640862 c.1941A>G p.R647R Chr.7: 81705459-81705459 on Build GRCh38
1623130 c.1984G>T p.E662* Chr.7: 81705416-81705416 on Build GRCh38
6952698 c.1948G>A p.V650M Chr.7: 81705452-81705452 on Build GRCh38
6427700 c.1946A>T p.K649M Chr.7: 81705454-81705454 on Build GRCh38
4153085 c.2000G>C p.G667A Chr.7: 81705400-81705400 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769580628 Chr.7: 81705419-81705419 on Build GRCh38
rs745642167 Chr.7: 81705382-81705383 on Build GRCh38
rs371548104 Chr.7: 81705344-81705344 on Build GRCh38
rs769761971 Chr.7: 81705237-81705237 on Build GRCh38
rs781715040 Chr.7: 81705361-81705361 on Build GRCh38
rs375394415 Chr.7: 81705351-81705351 on Build GRCh38
rs761664295 Chr.7: 81705410-81705410 on Build GRCh38
rs998501328 Chr.7: 81705318-81705321 on Build GRCh38
rs776576073 Chr.7: 81705406-81705406 on Build GRCh38
rs778031781 Chr.7: 81705378-81705378 on Build GRCh38
rs772967560 Chr.7: 81705421-81705421 on Build GRCh38
rs750048589 Chr.7: 81705350-81705350 on Build GRCh38
rs781145595 Chr.7: 81705342-81705342 on Build GRCh38
rs117001739 Chr.7: 81705277-81705277 on Build GRCh38
rs772105726 Chr.7: 81705393-81705393 on Build GRCh38
rs201730498 Chr.7: 81705381-81705381 on Build GRCh38
rs192932545 Chr.7: 81705369-81705369 on Build GRCh38
rs766007676 Chr.7: 81705435-81705435 on Build GRCh38
rs944137353 Chr.7: 81705275-81705275 on Build GRCh38
rs745942461 Chr.7: 81705384-81705384 on Build GRCh38
rs780620898 Chr.7: 81705333-81705333 on Build GRCh38
rs553489977 Chr.7: 81705354-81705354 on Build GRCh38
rs749474633 Chr.7: 81705380-81705380 on Build GRCh38
rs766064717 Chr.7: 81705353-81705353 on Build GRCh38
rs187442152 Chr.7: 81705335-81705335 on Build GRCh38
rs5745755 Chr.7: 81705332-81705332 on Build GRCh38
rs779318616 Chr.7: 81705382-81705382 on Build GRCh38
rs200169107 Chr.7: 81705359-81705359 on Build GRCh38
rs536246454 Chr.7: 81705366-81705366 on Build GRCh38

View Full Product Details