Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 148996689-148996962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GYG1
Entrez Gene ID
2992
Gene Name
glycogenin 1
Gene Aliases
GSD15, GYG
UniGene
Hs.477892
Gene Chromosome Location
Chr.3: 148991408-149027669 on Build GRCh38
Transcripts
NM_001184720.1 NM_001184721.1 NM_004130.3 XM_017006275.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7078932 c.402C>A p.S134S Chr.3: 148996825-148996825 on Build GRCh38
728810 c.380G>C p.G127A Chr.3: 148996803-148996803 on Build GRCh38
4114325 c.477T>C p.F159F Chr.3: 148996900-148996900 on Build GRCh38
6433809 c.416A>G p.Y139C Chr.3: 148996839-148996839 on Build GRCh38
4836121 c.334G>A p.D112N Chr.3: 148996757-148996757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs978877121 Chr.3: 148996771-148996771 on Build GRCh38
rs758175737 Chr.3: 148996742-148996742 on Build GRCh38
rs770259395 Chr.3: 148996765-148996765 on Build GRCh38
rs776240347 Chr.3: 148996825-148996825 on Build GRCh38
rs528341533 Chr.3: 148996861-148996861 on Build GRCh38
rs750209607 Chr.3: 148996737-148996737 on Build GRCh38
rs757811106 Chr.3: 148996857-148996873 on Build GRCh38
rs144099043 Chr.3: 148996723-148996723 on Build GRCh38
rs751667472 Chr.3: 148996752-148996752 on Build GRCh38
rs760183284 Chr.3: 148996811-148996811 on Build GRCh38
rs765243532 Chr.3: 148996835-148996835 on Build GRCh38
rs942145920 Chr.3: 148996880-148996880 on Build GRCh38
rs748347481 Chr.3: 148996904-148996904 on Build GRCh38
rs749686740 Chr.3: 148996935-148996935 on Build GRCh38
rs756594918 Chr.3: 148996842-148996842 on Build GRCh38
rs536913225 Chr.3: 148996920-148996920 on Build GRCh38
rs937188326 Chr.3: 148996834-148996834 on Build GRCh38
rs750396146 Chr.3: 148996852-148996852 on Build GRCh38
rs987032648 Chr.3: 148996802-148996802 on Build GRCh38
rs114156848 Chr.3: 148996730-148996730 on Build GRCh38
rs566956486 Chr.3: 148996826-148996826 on Build GRCh38
rs377349105 Chr.3: 148996923-148996923 on Build GRCh38
rs35054019 Chr.3: 148996875-148996875 on Build GRCh38
rs373007689 Chr.3: 148996817-148996817 on Build GRCh38
rs968503138 Chr.3: 148996760-148996760 on Build GRCh38
rs771263768 Chr.3: 148996799-148996799 on Build GRCh38
rs775059459 Chr.3: 148996714-148996714 on Build GRCh38
rs774778687 Chr.3: 148996807-148996807 on Build GRCh38
rs1028467140 Chr.3: 148996777-148996777 on Build GRCh38
rs1020360836 Chr.3: 148996758-148996758 on Build GRCh38
rs760225151 Chr.3: 148996718-148996718 on Build GRCh38
rs764225115 Chr.3: 148996729-148996731 on Build GRCh38
rs146500395 Chr.3: 148996727-148996727 on Build GRCh38
rs201376488 Chr.3: 148996922-148996922 on Build GRCh38
rs752756096 Chr.3: 148996908-148996908 on Build GRCh38
rs868642694 Chr.3: 148996883-148996883 on Build GRCh38
rs780320883 Chr.3: 148996744-148996744 on Build GRCh38
rs920037396 Chr.3: 148996902-148996902 on Build GRCh38
rs748262447 Chr.3: 148996763-148996763 on Build GRCh38
rs201036138 Chr.3: 148996786-148996786 on Build GRCh38
rs749654439 Chr.3: 148996779-148996779 on Build GRCh38
rs140175164 Chr.3: 148996862-148996862 on Build GRCh38
rs974043471 Chr.3: 148996896-148996896 on Build GRCh38
rs955833273 Chr.3: 148996790-148996790 on Build GRCh38
rs751394282 Chr.3: 148996872-148996872 on Build GRCh38
rs781200988 Chr.3: 148996762-148996762 on Build GRCh38
rs558790379 Chr.3: 148996871-148996871 on Build GRCh38
rs753901064 Chr.3: 148996772-148996773 on Build GRCh38
rs200687148 Chr.3: 148996755-148996755 on Build GRCh38

View Full Product Details