Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 100633058-100633313 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRG7
Entrez Gene ID
84873
Gene Name
adhesion G protein-coupled receptor G7
Gene Aliases
GPR128
UniGene
Hs.334511
Gene Chromosome Location
Chr.3: 100609589-100696174 on Build GRCh38
Transcripts
NM_032787.2 XM_011513245.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4545652 c.382G>A p.E128K Chr.3: 100633312-100633312 on Build GRCh38
6843098 c.355C>T p.R119W Chr.3: 100633285-100633285 on Build GRCh38
4489168 c.343C>T p.P115S Chr.3: 100633273-100633273 on Build GRCh38
3584523 c.358T>C p.L120L Chr.3: 100633288-100633288 on Build GRCh38
4892721 c.373C>T p.L125L Chr.3: 100633303-100633303 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752320101 Chr.3: 100633275-100633275 on Build GRCh38
rs769533373 Chr.3: 100633086-100633086 on Build GRCh38
rs199760799 Chr.3: 100633177-100633178 on Build GRCh38
rs1018456603 Chr.3: 100633145-100633145 on Build GRCh38
rs914539899 Chr.3: 100633252-100633252 on Build GRCh38
rs757832499 Chr.3: 100633276-100633276 on Build GRCh38
rs114672200 Chr.3: 100633266-100633266 on Build GRCh38
rs964886900 Chr.3: 100633171-100633171 on Build GRCh38
rs73145104 Chr.3: 100633097-100633097 on Build GRCh38
rs762565256 Chr.3: 100633242-100633242 on Build GRCh38
rs766311385 Chr.3: 100633260-100633260 on Build GRCh38
rs987468023 Chr.3: 100633142-100633142 on Build GRCh38
rs998119894 Chr.3: 100633198-100633198 on Build GRCh38
rs200895145 Chr.3: 100633265-100633265 on Build GRCh38
rs937168407 Chr.3: 100633227-100633227 on Build GRCh38
rs989024420 Chr.3: 100633211-100633211 on Build GRCh38
rs610854 Chr.3: 100633199-100633199 on Build GRCh38
rs531782420 Chr.3: 100633096-100633096 on Build GRCh38
rs369393091 Chr.3: 100633285-100633285 on Build GRCh38
rs753641282 Chr.3: 100633264-100633264 on Build GRCh38
rs1048130468 Chr.3: 100633091-100633091 on Build GRCh38
rs1006612155 Chr.3: 100633138-100633138 on Build GRCh38
rs143646459 Chr.3: 100633286-100633286 on Build GRCh38
rs973877778 Chr.3: 100633207-100633207 on Build GRCh38
rs1029239190 Chr.3: 100633092-100633092 on Build GRCh38
rs920718137 Chr.3: 100633225-100633225 on Build GRCh38
rs760591292 Chr.3: 100633253-100633253 on Build GRCh38
rs956327568 Chr.3: 100633204-100633204 on Build GRCh38
rs750308740 Chr.3: 100633249-100633249 on Build GRCh38

View Full Product Details