Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 101543116-101543383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRHL2
Entrez Gene ID
79977
Gene Name
grainyhead like transcription factor 2
Gene Aliases
BOM, DFNA28, ECTDS, TFCP2L3
UniGene
Hs.661088
Gene Chromosome Location
Chr.8: 101492440-101678105 on Build GRCh38
Transcripts
NM_024915.3 XM_011517305.2 XM_011517306.2 XM_011517307.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6179246 c.120G>T p.L40F Chr.8: 101543340-101543340 on Build GRCh38
4523553 c.121G>A p.E41K Chr.8: 101543341-101543341 on Build GRCh38
4523888 c.123G>A p.E41E Chr.8: 101543343-101543343 on Build GRCh38
281513 c.21-1G>T p.? Chr.8: 101543240-101543240 on Build GRCh38
3833753 c.76C>T p.R26* Chr.8: 101543296-101543296 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779951984 Chr.8: 101543301-101543301 on Build GRCh38
rs777303874 Chr.8: 101543229-101543229 on Build GRCh38
rs756952344 Chr.8: 101543275-101543275 on Build GRCh38
rs769228709 Chr.8: 101543349-101543349 on Build GRCh38
rs780810033 Chr.8: 101543307-101543307 on Build GRCh38
rs764407394 Chr.8: 101543274-101543274 on Build GRCh38
rs759826730 Chr.8: 101543260-101543260 on Build GRCh38
rs768277942 Chr.8: 101543219-101543219 on Build GRCh38
rs975342182 Chr.8: 101543166-101543166 on Build GRCh38
rs73701923 Chr.8: 101543217-101543217 on Build GRCh38
rs995483376 Chr.8: 101543294-101543294 on Build GRCh38
rs898759692 Chr.8: 101543281-101543281 on Build GRCh38
rs1052546011 Chr.8: 101543195-101543195 on Build GRCh38
rs749906501 Chr.8: 101543254-101543254 on Build GRCh38
rs1001767700 Chr.8: 101543253-101543253 on Build GRCh38
rs913630702 Chr.8: 101543199-101543199 on Build GRCh38
rs778995557 Chr.8: 101543292-101543292 on Build GRCh38
rs1030232476 Chr.8: 101543165-101543165 on Build GRCh38
rs753662454 Chr.8: 101543215-101543215 on Build GRCh38
rs747671817 Chr.8: 101543338-101543338 on Build GRCh38
rs776162436 Chr.8: 101543221-101543221 on Build GRCh38
rs774784244 Chr.8: 101543259-101543259 on Build GRCh38
rs752980201 Chr.8: 101543269-101543269 on Build GRCh38
rs760997445 Chr.8: 101543270-101543270 on Build GRCh38
rs779596660 Chr.8: 101543296-101543296 on Build GRCh38
rs374676730 Chr.8: 101543223-101543223 on Build GRCh38
rs778497540 Chr.8: 101543289-101543289 on Build GRCh38
rs557545847 Chr.8: 101543141-101543141 on Build GRCh38
rs200212977 Chr.8: 101543237-101543237 on Build GRCh38
rs3735709 Chr.8: 101543246-101543246 on Build GRCh38
rs912391872 Chr.8: 101543144-101543144 on Build GRCh38
rs754025898 Chr.8: 101543285-101543285 on Build GRCh38
rs993904280 Chr.8: 101543356-101543356 on Build GRCh38
rs758153213 Chr.8: 101543300-101543300 on Build GRCh38
rs767872185 Chr.8: 101543265-101543265 on Build GRCh38
rs1018031449 Chr.8: 101543228-101543228 on Build GRCh38
rs890388714 Chr.8: 101543184-101543184 on Build GRCh38
rs561693958 Chr.8: 101543303-101543303 on Build GRCh38
rs750332552 Chr.8: 101543297-101543297 on Build GRCh38

View Full Product Details