Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 38982393-38982664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTRA4
Entrez Gene ID
203100
Gene Name
HtrA serine peptidase 4
Gene Aliases
-
UniGene
Hs.661014
Gene Chromosome Location
Chr.8: 38974149-38988662 on Build GRCh38
Transcripts
NM_153692.3 XM_011544431.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3367347 c.1115G>T p.G372V Chr.8: 38982498-38982498 on Build GRCh38
1239106 c.1157T>C p.L386P Chr.8: 38982540-38982540 on Build GRCh38
3900092 c.1167T>C p.T389T Chr.8: 38982550-38982550 on Build GRCh38
5874670 c.1120G>A p.A374T Chr.8: 38982503-38982503 on Build GRCh38
3951654 c.1170G>T p.V390V Chr.8: 38982553-38982553 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1010822790 Chr.8: 38982437-38982437 on Build GRCh38
rs752595969 Chr.8: 38982487-38982487 on Build GRCh38
rs374833705 Chr.8: 38982465-38982465 on Build GRCh38
rs770040324 Chr.8: 38982555-38982555 on Build GRCh38
rs745411832 Chr.8: 38982522-38982522 on Build GRCh38
rs769004292 Chr.8: 38982536-38982536 on Build GRCh38
rs759435415 Chr.8: 38982481-38982481 on Build GRCh38
rs777434731 Chr.8: 38982504-38982504 on Build GRCh38
rs918660807 Chr.8: 38982507-38982507 on Build GRCh38
rs756090982 Chr.8: 38982462-38982462 on Build GRCh38
rs572734830 Chr.8: 38982448-38982448 on Build GRCh38
rs778760581 Chr.8: 38982598-38982598 on Build GRCh38
rs929451966 Chr.8: 38982619-38982619 on Build GRCh38
rs370158891 Chr.8: 38982604-38982604 on Build GRCh38
rs148796117 Chr.8: 38982528-38982528 on Build GRCh38
rs372702750 Chr.8: 38982572-38982572 on Build GRCh38
rs200095456 Chr.8: 38982545-38982545 on Build GRCh38
rs560757151 Chr.8: 38982516-38982516 on Build GRCh38
rs750692046 Chr.8: 38982474-38982475 on Build GRCh38
rs965346875 Chr.8: 38982482-38982483 on Build GRCh38
rs778564119 Chr.8: 38982517-38982517 on Build GRCh38
rs763037727 Chr.8: 38982563-38982563 on Build GRCh38
rs777033515 Chr.8: 38982537-38982537 on Build GRCh38
rs370712609 Chr.8: 38982447-38982447 on Build GRCh38
rs116073133 Chr.8: 38982556-38982556 on Build GRCh38
rs756941127 Chr.8: 38982595-38982595 on Build GRCh38
rs772438569 Chr.8: 38982523-38982523 on Build GRCh38
rs112522696 Chr.8: 38982623-38982623 on Build GRCh38
rs367814294 Chr.8: 38982480-38982480 on Build GRCh38
rs1047480705 Chr.8: 38982636-38982636 on Build GRCh38
rs182946954 Chr.8: 38982585-38982585 on Build GRCh38
rs376517015 Chr.8: 38982606-38982606 on Build GRCh38
rs375232321 Chr.8: 38982449-38982449 on Build GRCh38
rs755897562 Chr.8: 38982503-38982503 on Build GRCh38
rs774086219 Chr.8: 38982612-38982613 on Build GRCh38
rs373659628 Chr.8: 38982588-38982588 on Build GRCh38
rs752612644 Chr.8: 38982577-38982577 on Build GRCh38
rs767538340 Chr.8: 38982486-38982486 on Build GRCh38
rs551752587 Chr.8: 38982505-38982505 on Build GRCh38
rs780564229 Chr.8: 38982525-38982525 on Build GRCh38
rs758634845 Chr.8: 38982590-38982590 on Build GRCh38
rs954101757 Chr.8: 38982412-38982413 on Build GRCh38
rs985361281 Chr.8: 38982443-38982443 on Build GRCh38

View Full Product Details