Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 31924399-31924672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HUNK
Entrez Gene ID
30811
Gene Name
hormonally up-regulated Neu-associated kinase
Gene Aliases
-
UniGene
Hs.109437
Gene Chromosome Location
Chr.21: 31873315-32004064 on Build GRCh38
Transcripts
NM_014586.1 XM_011529537.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3389972 c.356G>A p.R119H Chr.21: 31924562-31924562 on Build GRCh38
6683152 c.432_433insG p.N147fs*8 Chr.21: 31924638-31924639 on Build GRCh38
1413738 c.328C>T p.R110* Chr.21: 31924534-31924534 on Build GRCh38
1030104 c.453G>T p.K151N Chr.21: 31924659-31924659 on Build GRCh38
5680731 c.335G>A p.G112D Chr.21: 31924541-31924541 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772179174 Chr.21: 31924554-31924554 on Build GRCh38
rs564370055 Chr.21: 31924620-31924620 on Build GRCh38
rs749326655 Chr.21: 31924421-31924421 on Build GRCh38
rs756689144 Chr.21: 31924482-31924482 on Build GRCh38
rs374827442 Chr.21: 31924439-31924439 on Build GRCh38
rs760731052 Chr.21: 31924457-31924457 on Build GRCh38
rs544212060 Chr.21: 31924563-31924563 on Build GRCh38
rs758015472 Chr.21: 31924499-31924499 on Build GRCh38
rs763925469 Chr.21: 31924465-31924465 on Build GRCh38
rs749932370 Chr.21: 31924610-31924610 on Build GRCh38
rs753113608 Chr.21: 31924641-31924641 on Build GRCh38
rs199631073 Chr.21: 31924440-31924440 on Build GRCh38
rs779109858 Chr.21: 31924532-31924532 on Build GRCh38
rs993341188 Chr.21: 31924420-31924420 on Build GRCh38
rs778078613 Chr.21: 31924516-31924516 on Build GRCh38
rs746129084 Chr.21: 31924553-31924553 on Build GRCh38
rs768998000 Chr.21: 31924566-31924566 on Build GRCh38
rs545460097 Chr.21: 31924520-31924520 on Build GRCh38
rs761716649 Chr.21: 31924601-31924601 on Build GRCh38
rs977013844 Chr.21: 31924481-31924481 on Build GRCh38
rs756660257 Chr.21: 31924515-31924515 on Build GRCh38
rs201964145 Chr.21: 31924455-31924455 on Build GRCh38
rs55847614 Chr.21: 31924638-31924638 on Build GRCh38
rs752994349 Chr.21: 31924510-31924510 on Build GRCh38
rs201574800 Chr.21: 31924445-31924445 on Build GRCh38
rs753357737 Chr.21: 31924474-31924474 on Build GRCh38
rs760328005 Chr.21: 31924562-31924562 on Build GRCh38
rs138361949 Chr.21: 31924493-31924493 on Build GRCh38
rs142458503 Chr.21: 31924531-31924531 on Build GRCh38
rs914540613 Chr.21: 31924629-31924629 on Build GRCh38
rs781669281 Chr.21: 31924506-31924506 on Build GRCh38
rs775412939 Chr.21: 31924561-31924561 on Build GRCh38
rs776504720 Chr.21: 31924590-31924590 on Build GRCh38
rs146277869 Chr.21: 31924602-31924602 on Build GRCh38
rs148302130 Chr.21: 31924632-31924632 on Build GRCh38
rs775112603 Chr.21: 31924452-31924452 on Build GRCh38
rs750127108 Chr.21: 31924498-31924498 on Build GRCh38

View Full Product Details