Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35618175-35618434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAUS5
Entrez Gene ID
23354
Gene Name
HAUS augmin like complex subunit 5
Gene Aliases
KIAA0841, dgt5
UniGene
Hs.7426
Gene Chromosome Location
Chr.19: 35612744-35625349 on Build GRCh38
Transcripts
NM_015302.1 XM_011526684.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6377661 c.827A>C p.Q276P Chr.19: 35618407-35618407 on Build GRCh38
7230263 c.830C>A p.A277D Chr.19: 35618410-35618410 on Build GRCh38
4567709 c.834_835GG>CA p.D279N Chr.19: 35618414-35618415 on Build GRCh38
6384933 c.839A>T p.Q280L Chr.19: 35618419-35618419 on Build GRCh38
6382587 c.823C>A p.P275T Chr.19: 35618403-35618403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777107012 Chr.19: 35618368-35618368 on Build GRCh38
rs896361693 Chr.19: 35618346-35618346 on Build GRCh38
rs745717731 Chr.19: 35618212-35618212 on Build GRCh38
rs370240829 Chr.19: 35618411-35618411 on Build GRCh38
rs757265956 Chr.19: 35618208-35618208 on Build GRCh38
rs746462570 Chr.19: 35618207-35618207 on Build GRCh38
rs772775791 Chr.19: 35618367-35618367 on Build GRCh38
rs372188303 Chr.19: 35618363-35618363 on Build GRCh38
rs748346602 Chr.19: 35618241-35618241 on Build GRCh38
rs758000179 Chr.19: 35618387-35618387 on Build GRCh38
rs753058003 Chr.19: 35618375-35618375 on Build GRCh38
rs1056539694 Chr.19: 35618345-35618345 on Build GRCh38
rs773290699 Chr.19: 35618357-35618357 on Build GRCh38
rs923605758 Chr.19: 35618336-35618336 on Build GRCh38
rs367779153 Chr.19: 35618371-35618371 on Build GRCh38
rs550932004 Chr.19: 35618315-35618315 on Build GRCh38
rs1043436795 Chr.19: 35618203-35618203 on Build GRCh38
rs1026891321 Chr.19: 35618204-35618204 on Build GRCh38
rs1015550456 Chr.19: 35618361-35618361 on Build GRCh38
rs779731836 Chr.19: 35618414-35618414 on Build GRCh38
rs1001846921 Chr.19: 35618261-35618261 on Build GRCh38
rs769458606 Chr.19: 35618220-35618220 on Build GRCh38
rs2285412 Chr.19: 35618410-35618410 on Build GRCh38
rs959187708 Chr.19: 35618219-35618219 on Build GRCh38
rs977751556 Chr.19: 35618289-35618289 on Build GRCh38
rs530974920 Chr.19: 35618293-35618293 on Build GRCh38
rs904956885 Chr.19: 35618226-35618226 on Build GRCh38
rs771071069 Chr.19: 35618195-35618195 on Build GRCh38
rs756035655 Chr.19: 35618413-35618413 on Build GRCh38
rs368036069 Chr.19: 35618228-35618228 on Build GRCh38
rs375254861 Chr.19: 35618370-35618370 on Build GRCh38
rs781273302 Chr.19: 35618210-35618210 on Build GRCh38
rs763737522 Chr.19: 35618389-35618389 on Build GRCh38
rs374135452 Chr.19: 35618221-35618221 on Build GRCh38
rs772284095 Chr.19: 35618355-35618355 on Build GRCh38
rs751184887 Chr.19: 35618394-35618394 on Build GRCh38
rs774678464 Chr.19: 35618233-35618233 on Build GRCh38
rs970899194 Chr.19: 35618285-35618285 on Build GRCh38
rs567485439 Chr.19: 35618364-35618364 on Build GRCh38
rs777170720 Chr.19: 35618198-35618198 on Build GRCh38

View Full Product Details