Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 141780237-141780502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRK7
Entrez Gene ID
131890
Gene Name
G protein-coupled receptor kinase 7
Gene Aliases
GPRK7
UniGene
Hs.680654
Gene Chromosome Location
Chr.3: 141763127-141817373 on Build GRCh38
Transcripts
NM_139209.2 XM_017005717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
48870 c.733A>T p.K245* Chr.3: 141780494-141780494 on Build GRCh38
1419587 c.726C>A p.I242I Chr.3: 141780487-141780487 on Build GRCh38
6675814 c.622G>A p.V208I Chr.3: 141780383-141780383 on Build GRCh38
7255364 c.682_689delAAGAAGAA p.K228fs*29 Chr.3: 141780443-141780450 on Build GRCh38
5059370 c.741C>T p.S247S Chr.3: 141780502-141780502 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550144618 Chr.3: 141780383-141780383 on Build GRCh38
rs746903414 Chr.3: 141780362-141780362 on Build GRCh38
rs974495658 Chr.3: 141780463-141780463 on Build GRCh38
rs749651118 Chr.3: 141780407-141780407 on Build GRCh38
rs781303819 Chr.3: 141780381-141780381 on Build GRCh38
rs1006305074 Chr.3: 141780265-141780265 on Build GRCh38
rs758207753 Chr.3: 141780457-141780457 on Build GRCh38
rs779975659 Chr.3: 141780359-141780359 on Build GRCh38
rs748030827 Chr.3: 141780436-141780452 on Build GRCh38
rs751163213 Chr.3: 141780343-141780343 on Build GRCh38
rs780990873 Chr.3: 141780351-141780353 on Build GRCh38
rs757594701 Chr.3: 141780416-141780416 on Build GRCh38
rs770587889 Chr.3: 141780318-141780318 on Build GRCh38
rs139715496 Chr.3: 141780438-141780438 on Build GRCh38
rs771364879 Chr.3: 141780413-141780413 on Build GRCh38
rs764997117 Chr.3: 141780337-141780337 on Build GRCh38
rs555247174 Chr.3: 141780296-141780298 on Build GRCh38
rs142906049 Chr.3: 141780344-141780344 on Build GRCh38
rs763560113 Chr.3: 141780435-141780435 on Build GRCh38
rs892889836 Chr.3: 141780374-141780374 on Build GRCh38
rs35566288 Chr.3: 141780437-141780437 on Build GRCh38
rs372254837 Chr.3: 141780467-141780467 on Build GRCh38
rs185026248 Chr.3: 141780336-141780336 on Build GRCh38
rs1037751620 Chr.3: 141780274-141780274 on Build GRCh38
rs773517151 Chr.3: 141780399-141780399 on Build GRCh38
rs749981895 Chr.3: 141780456-141780456 on Build GRCh38
rs878945266 Chr.3: 141780302-141780302 on Build GRCh38
rs749675735 Chr.3: 141780408-141780408 on Build GRCh38
rs754889750 Chr.3: 141780468-141780468 on Build GRCh38
rs546001827 Chr.3: 141780329-141780329 on Build GRCh38
rs755084843 Chr.3: 141780370-141780370 on Build GRCh38
rs147462832 Chr.3: 141780427-141780427 on Build GRCh38
rs192098472 Chr.3: 141780287-141780287 on Build GRCh38
rs368279239 Chr.3: 141780382-141780382 on Build GRCh38
rs960298425 Chr.3: 141780346-141780346 on Build GRCh38
rs369552626 Chr.3: 141780458-141780458 on Build GRCh38
rs758119429 Chr.3: 141780345-141780345 on Build GRCh38

View Full Product Details