Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 41695986-41696241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASK
Entrez Gene ID
8573
Gene Name
calcium/calmodulin dependent serine protein kinase
Gene Aliases
CAGH39, CAMGUK, CMG, FGS4, LIN2, MICPCH, MRXSNA, TNRC8
UniGene
Hs.495984
Gene Chromosome Location
Chr.X: 41514934-41923525 on Build GRCh38
Transcripts
NM_001126054.2 NM_001126055.2 NM_003688.3 XM_005272686.4 XM_006724566.3 XM_011543993.2 XM_011543994.2 XM_011543995.2 XM_011543996.2 NM_001097579.1 NM_005300.3 XM_005272597.3

Gene Symbol
GPR34
Entrez Gene ID
2857
Gene Name
G protein-coupled receptor 34
Gene Aliases
LYPSR1
UniGene
Hs.495989
Gene Chromosome Location
Chr.X: 41686543-41697277 on Build GRCh38
Transcripts
NM_001126054.2 NM_001126055.2 NM_003688.3 XM_005272686.4 XM_006724566.3 XM_011543993.2 XM_011543994.2 XM_011543995.2 XM_011543996.2 NM_001097579.1 NM_005300.3 XM_005272597.3

SNP Detail

SNP ID Chromosome Location
rs753126394 Chr.X: 41696154-41696154 on Build GRCh38
rs895214157 Chr.X: 41696172-41696172 on Build GRCh38
rs746916394 Chr.X: 41696105-41696105 on Build GRCh38
rs370941111 Chr.X: 41696053-41696053 on Build GRCh38
rs373704509 Chr.X: 41696152-41696152 on Build GRCh38
rs772869981 Chr.X: 41696130-41696130 on Build GRCh38
rs768784541 Chr.X: 41696106-41696106 on Build GRCh38
rs1016302316 Chr.X: 41696046-41696046 on Build GRCh38
rs769217317 Chr.X: 41696121-41696121 on Build GRCh38
rs1006633242 Chr.X: 41696040-41696040 on Build GRCh38
rs761156570 Chr.X: 41696160-41696160 on Build GRCh38
rs754303572 Chr.X: 41696199-41696199 on Build GRCh38
rs771196302 Chr.X: 41696031-41696032 on Build GRCh38
rs1013685174 Chr.X: 41696201-41696201 on Build GRCh38
rs1000860910 Chr.X: 41696087-41696087 on Build GRCh38
rs762559517 Chr.X: 41696145-41696145 on Build GRCh38
rs759763316 Chr.X: 41696194-41696194 on Build GRCh38
rs757321606 Chr.X: 41696204-41696204 on Build GRCh38
rs761332593 Chr.X: 41696113-41696113 on Build GRCh38
rs771945164 Chr.X: 41696051-41696051 on Build GRCh38
rs764525670 Chr.X: 41696169-41696169 on Build GRCh38

View Full Product Details