Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6381460-6381717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2F1
Entrez Gene ID
2962
Gene Name
general transcription factor IIF subunit 1
Gene Aliases
BTF4, RAP74, TF2F1, TFIIF
UniGene
Hs.68257
Gene Chromosome Location
Chr.19: 6379569-6393280 on Build GRCh38
Transcripts
NM_002096.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5755505 c.904G>A p.D302N Chr.19: 6381473-6381473 on Build GRCh38
7271938 c.889G>T p.G297W Chr.19: 6381563-6381563 on Build GRCh38
4942697 c.879G>A p.Q293Q Chr.19: 6381573-6381573 on Build GRCh38
1002876 c.887_889delAGG p.E296delE Chr.19: 6381563-6381565 on Build GRCh38
5889369 c.899-4C>T p.? Chr.19: 6381482-6381482 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368420170 Chr.19: 6381532-6381532 on Build GRCh38
rs747943929 Chr.19: 6381635-6381635 on Build GRCh38
rs529038407 Chr.19: 6381528-6381528 on Build GRCh38
rs201186081 Chr.19: 6381486-6381486 on Build GRCh38
rs7247287 Chr.19: 6381664-6381664 on Build GRCh38
rs772764070 Chr.19: 6381576-6381576 on Build GRCh38
rs375015426 Chr.19: 6381493-6381493 on Build GRCh38
rs753039298 Chr.19: 6381600-6381600 on Build GRCh38
rs777874021 Chr.19: 6381541-6381541 on Build GRCh38
rs200161164 Chr.19: 6381646-6381646 on Build GRCh38
rs368166573 Chr.19: 6381673-6381673 on Build GRCh38
rs376212238 Chr.19: 6381663-6381663 on Build GRCh38
rs891318943 Chr.19: 6381508-6381508 on Build GRCh38
rs370742482 Chr.19: 6381686-6381686 on Build GRCh38
rs368496125 Chr.19: 6381677-6381677 on Build GRCh38
rs78149805 Chr.19: 6381689-6381689 on Build GRCh38
rs376228766 Chr.19: 6381650-6381650 on Build GRCh38
rs767420292 Chr.19: 6381666-6381666 on Build GRCh38
rs112970874 Chr.19: 6381652-6381652 on Build GRCh38
rs780872673 Chr.19: 6381570-6381570 on Build GRCh38
rs368799788 Chr.19: 6381618-6381618 on Build GRCh38
rs775707314 Chr.19: 6381492-6381492 on Build GRCh38
rs773987502 Chr.19: 6381533-6381534 on Build GRCh38
rs752865503 Chr.19: 6381490-6381490 on Build GRCh38
rs113337492 Chr.19: 6381626-6381626 on Build GRCh38
rs916305909 Chr.19: 6381485-6381485 on Build GRCh38
rs140250398 Chr.19: 6381679-6381679 on Build GRCh38
rs1017730863 Chr.19: 6381684-6381684 on Build GRCh38
rs781627280 Chr.19: 6381488-6381488 on Build GRCh38
rs367852832 Chr.19: 6381514-6381514 on Build GRCh38
rs770567679 Chr.19: 6381579-6381579 on Build GRCh38
rs376177994 Chr.19: 6381536-6381537 on Build GRCh38
rs147542094 Chr.19: 6381525-6381525 on Build GRCh38
rs765317586 Chr.19: 6381624-6381624 on Build GRCh38
rs772629783 Chr.19: 6381523-6381523 on Build GRCh38
rs769300843 Chr.19: 6381522-6381522 on Build GRCh38
rs760925148 Chr.19: 6381603-6381603 on Build GRCh38
rs182233096 Chr.19: 6381643-6381643 on Build GRCh38
rs746342055 Chr.19: 6381648-6381648 on Build GRCh38
rs753918082 Chr.19: 6381547-6381547 on Build GRCh38
rs373905184 Chr.19: 6381672-6381672 on Build GRCh38
rs369353212 Chr.19: 6381647-6381647 on Build GRCh38
rs778541739 Chr.19: 6381678-6381678 on Build GRCh38
rs745774284 Chr.19: 6381496-6381496 on Build GRCh38
rs371724007 Chr.19: 6381515-6381515 on Build GRCh38
rs201495032 Chr.19: 6381685-6381685 on Build GRCh38
rs200767408 Chr.19: 6381628-6381628 on Build GRCh38
rs765262160 Chr.19: 6381655-6381655 on Build GRCh38
rs979171881 Chr.19: 6381627-6381627 on Build GRCh38
rs751967759 Chr.19: 6381531-6381531 on Build GRCh38
rs372036105 Chr.19: 6381536-6381536 on Build GRCh38
rs923553778 Chr.19: 6381582-6381582 on Build GRCh38
rs752018799 Chr.19: 6381479-6381479 on Build GRCh38
rs369703284 Chr.19: 6381526-6381526 on Build GRCh38
rs10425395 Chr.19: 6381649-6381649 on Build GRCh38
rs746150695 Chr.19: 6381478-6381478 on Build GRCh38
rs200800402 Chr.19: 6381575-6381575 on Build GRCh38
rs746955897 Chr.19: 6381556-6381556 on Build GRCh38
rs747908686 Chr.19: 6381643-6381644 on Build GRCh38
rs139987959 Chr.19: 6381617-6381617 on Build GRCh38
rs375041963 Chr.19: 6381517-6381517 on Build GRCh38
rs756318990 Chr.19: 6381537-6381537 on Build GRCh38
rs755785989 Chr.19: 6381641-6381641 on Build GRCh38
rs774939372 Chr.19: 6381580-6381580 on Build GRCh38
rs988440410 Chr.19: 6381612-6381612 on Build GRCh38
rs747797295 Chr.19: 6381572-6381572 on Build GRCh38
rs374972528 Chr.19: 6381644-6381644 on Build GRCh38
rs762618865 Chr.19: 6381577-6381577 on Build GRCh38
rs1051320129 Chr.19: 6381497-6381497 on Build GRCh38
rs776663368 Chr.19: 6381653-6381653 on Build GRCh38
rs780035055 Chr.19: 6381554-6381554 on Build GRCh38
rs768383952 Chr.19: 6381559-6381559 on Build GRCh38
rs374302740 Chr.19: 6381688-6381688 on Build GRCh38
rs761386657 Chr.19: 6381518-6381518 on Build GRCh38
rs8111068 Chr.19: 6381487-6381487 on Build GRCh38
rs767049604 Chr.19: 6381528-6381528 on Build GRCh38
rs758322168 Chr.19: 6381548-6381548 on Build GRCh38
rs772033811 Chr.19: 6381504-6381504 on Build GRCh38
rs376967252 Chr.19: 6381625-6381625 on Build GRCh38
rs201433302 Chr.19: 6381510-6381510 on Build GRCh38
rs375396180 Chr.19: 6381592-6381592 on Build GRCh38
rs760139557 Chr.19: 6381590-6381590 on Build GRCh38
rs531762713 Chr.19: 6381599-6381599 on Build GRCh38

View Full Product Details