Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 96832660-96832901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HS6ST3
Entrez Gene ID
266722
Gene Name
heparan sulfate 6-O-sulfotransferase 3
Gene Aliases
HS6ST-3
UniGene
Hs.171001
Gene Chromosome Location
Chr.13: 96090634-96839562 on Build GRCh38
Transcripts
NM_153456.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3943291 c.989A>G p.E330G Chr.13: 96832771-96832771 on Build GRCh38
469631 c.908A>G p.Y303C Chr.13: 96832690-96832690 on Build GRCh38
1210013 c.1052T>C p.F351S Chr.13: 96832834-96832834 on Build GRCh38
77865 c.958G>A p.V320M Chr.13: 96832740-96832740 on Build GRCh38
3885565 c.1009C>T p.L337L Chr.13: 96832791-96832791 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751137557 Chr.13: 96832771-96832771 on Build GRCh38
rs1010248410 Chr.13: 96832793-96832793 on Build GRCh38
rs565845833 Chr.13: 96832862-96832862 on Build GRCh38
rs759719846 Chr.13: 96832760-96832760 on Build GRCh38
rs763499432 Chr.13: 96832708-96832708 on Build GRCh38
rs866128801 Chr.13: 96832798-96832798 on Build GRCh38
rs747297518 Chr.13: 96832716-96832716 on Build GRCh38
rs375298542 Chr.13: 96832685-96832685 on Build GRCh38
rs1001089895 Chr.13: 96832694-96832694 on Build GRCh38
rs765198671 Chr.13: 96832764-96832764 on Build GRCh38
rs868456363 Chr.13: 96832687-96832687 on Build GRCh38
rs531254937 Chr.13: 96832769-96832769 on Build GRCh38
rs761573907 Chr.13: 96832820-96832820 on Build GRCh38
rs755638285 Chr.13: 96832853-96832853 on Build GRCh38
rs767109653 Chr.13: 96832824-96832824 on Build GRCh38
rs144389562 Chr.13: 96832707-96832707 on Build GRCh38
rs770895513 Chr.13: 96832717-96832717 on Build GRCh38
rs1054566348 Chr.13: 96832705-96832705 on Build GRCh38
rs772545385 Chr.13: 96832681-96832681 on Build GRCh38
rs535937797 Chr.13: 96832733-96832733 on Build GRCh38
rs147857247 Chr.13: 96832838-96832838 on Build GRCh38

View Full Product Details