Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 137030884-137031111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR101
Entrez Gene ID
83550
Gene Name
G protein-coupled receptor 101
Gene Aliases
GPCR6, PAGH2
UniGene
Hs.350569
Gene Chromosome Location
Chr.X: 137030148-137031674 on Build GRCh38
Transcripts
NM_054021.1

SNP Detail

SNP ID Chromosome Location
rs1005833739 Chr.X: 137030969-137030969 on Build GRCh38
rs776749651 Chr.X: 137031045-137031045 on Build GRCh38
rs988027680 Chr.X: 137030919-137030919 on Build GRCh38
rs765180208 Chr.X: 137031081-137031081 on Build GRCh38
rs775145092 Chr.X: 137031083-137031083 on Build GRCh38
rs900943965 Chr.X: 137030909-137030909 on Build GRCh38
rs112447740 Chr.X: 137030940-137030940 on Build GRCh38
rs781683243 Chr.X: 137031000-137031000 on Build GRCh38
rs764803089 Chr.X: 137031016-137031016 on Build GRCh38
rs570565842 Chr.X: 137030915-137030915 on Build GRCh38
rs201779657 Chr.X: 137031008-137031008 on Build GRCh38
rs761882471 Chr.X: 137030907-137030907 on Build GRCh38
rs748195497 Chr.X: 137031027-137031027 on Build GRCh38
rs943423745 Chr.X: 137030977-137030977 on Build GRCh38
rs774287814 Chr.X: 137030904-137030904 on Build GRCh38
rs763729290 Chr.X: 137030990-137030990 on Build GRCh38
rs772237606 Chr.X: 137031039-137031039 on Build GRCh38
rs746338884 Chr.X: 137031002-137031002 on Build GRCh38
rs774570365 Chr.X: 137031017-137031017 on Build GRCh38
rs760824201 Chr.X: 137030938-137030955 on Build GRCh38
rs371482705 Chr.X: 137030998-137030998 on Build GRCh38
rs913755486 Chr.X: 137030922-137030922 on Build GRCh38
rs762744638 Chr.X: 137031090-137031090 on Build GRCh38
rs759521947 Chr.X: 137031072-137031072 on Build GRCh38
rs928002784 Chr.X: 137030953-137030953 on Build GRCh38
rs968247978 Chr.X: 137030928-137030928 on Build GRCh38
rs776660370 Chr.X: 137030916-137030916 on Build GRCh38
rs752442726 Chr.X: 137030966-137030966 on Build GRCh38
rs749588022 Chr.X: 137031012-137031012 on Build GRCh38
rs368827016 Chr.X: 137030991-137030991 on Build GRCh38
rs757975557 Chr.X: 137030976-137030976 on Build GRCh38
rs780378576 Chr.X: 137031009-137031009 on Build GRCh38
rs139111924 Chr.X: 137030963-137030963 on Build GRCh38
rs747548759 Chr.X: 137030929-137030929 on Build GRCh38

View Full Product Details