Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 188483358-188483589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GULP1
Entrez Gene ID
51454
Gene Name
GULP, engulfment adaptor PTB domain containing 1
Gene Aliases
CED-6, CED6, GULP
UniGene
Hs.470887
Gene Chromosome Location
Chr.2: 188291669-188595926 on Build GRCh38
Transcripts
NM_001252668.1 NM_001252669.1 NM_016315.3 NR_045562.1 NR_045563.1 XM_006712580.3 XM_006712581.1 XM_006712582.1 XM_006712583.3 XM_006712584.3 XM_006712585.1 XM_006712589.3 XM_011511329.1 XM_011511331.1 XM_011511332.1 XM_011511333.1 XM_011511335.2 XM_017004303.1 XM_017004304.1 XM_017004305.1 XM_017004306.1 XM_017004307.1 XM_017004308.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4996169 c.41T>C p.M14T Chr.2: 188483443-188483443 on Build GRCh38
5058512 c.42G>A p.M14I Chr.2: 188483444-188483444 on Build GRCh38
5045192 c.85G>T p.A29S Chr.2: 188483487-188483487 on Build GRCh38
5572070 c.39G>A p.W13* Chr.2: 188483441-188483441 on Build GRCh38
5934540 c.49C>T p.P17S Chr.2: 188483451-188483451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762560085 Chr.2: 188483476-188483476 on Build GRCh38
rs952938145 Chr.2: 188483560-188483560 on Build GRCh38
rs767268022 Chr.2: 188483468-188483468 on Build GRCh38
rs767922509 Chr.2: 188483405-188483405 on Build GRCh38
rs778908469 Chr.2: 188483403-188483403 on Build GRCh38
rs754207124 Chr.2: 188483534-188483534 on Build GRCh38
rs75821338 Chr.2: 188483547-188483547 on Build GRCh38
rs896628200 Chr.2: 188483475-188483475 on Build GRCh38
rs754070483 Chr.2: 188483393-188483393 on Build GRCh38
rs761256116 Chr.2: 188483500-188483500 on Build GRCh38
rs746937734 Chr.2: 188483441-188483441 on Build GRCh38
rs1013692102 Chr.2: 188483533-188483533 on Build GRCh38
rs142155437 Chr.2: 188483495-188483495 on Build GRCh38
rs545140276 Chr.2: 188483451-188483451 on Build GRCh38
rs764406391 Chr.2: 188483513-188483513 on Build GRCh38
rs756558812 Chr.2: 188483387-188483387 on Build GRCh38
rs758307259 Chr.2: 188483511-188483514 on Build GRCh38
rs375441419 Chr.2: 188483411-188483411 on Build GRCh38
rs577612121 Chr.2: 188483415-188483415 on Build GRCh38
rs867691291 Chr.2: 188483479-188483479 on Build GRCh38
rs778183430 Chr.2: 188483389-188483389 on Build GRCh38
rs765927248 Chr.2: 188483478-188483478 on Build GRCh38
rs1025077490 Chr.2: 188483418-188483418 on Build GRCh38
rs757470456 Chr.2: 188483398-188483398 on Build GRCh38
rs769511591 Chr.2: 188483474-188483474 on Build GRCh38
rs560393029 Chr.2: 188483457-188483457 on Build GRCh38
rs757513262 Chr.2: 188483543-188483543 on Build GRCh38
rs985232440 Chr.2: 188483555-188483555 on Build GRCh38
rs779981795 Chr.2: 188483420-188483420 on Build GRCh38
rs768506343 Chr.2: 188483449-188483449 on Build GRCh38

View Full Product Details