Gene Transcripts

Gene Symbol
Phf1
Entrez Gene ID
21652
Gene Name
PHD finger protein 1
Gene Aliases
AW557215, D17Ertd455e, Pcl1, Phf2, Tctex3
Chromosome Location
Chr. 17: 26933127 - 26937908 on Build GRCm38
UniGene ID
Mm.258953
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB011550.1 - 6 673
AK076365.1 - 4 1589
AK133474.1 - 6 641
AK139088.1 - 6 879
AK144793.1 - 6 857
AK155268.1 - 4 1618
BC015077.1 - 6 556
BC145595.1 - 5 647
BC145596.1 - 6 562
U81490.1 - 6 613

Gene Symbol
Cuta
Entrez Gene ID
67675
Gene Name
cutA divalent cation tolerance homolog (E. coli)
Gene Aliases
0610039D01Rik, 1810022E02Rik, 1810060C03Rik, 2700094G22Rik, AI326454
Chromosome Location
Chr. 17: 26937972 - 26939815 on Build GRCm38
UniGene ID
Mm.439919
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK145185.1 - 1301

View Full Product Details