Gene Transcripts

Gene Symbol
Tollip
Entrez Gene ID
54473
Gene Name
toll interacting protein
Gene Aliases
4930403G24Rik, 4931428G15Rik
Chromosome Location
Chr. 7: 141881582 - 141902465 on Build GRCm38
UniGene ID
Mm.103551
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023764.3 NP_076253.1 6 845
GenBank mRNA AJ242971.1 - 6 849
AK015062.1 - 788
AK016482.1 - 6 846
AK045422.1 - 6 829
AK049263.1 - 6 867
AK049394.1 - 6 791
AK161312.1 - 6 832
BC053515.1 - 6 832
BC062139.1 - 6 826 6334946

View Full Product Details