Gene Transcripts

Gene Symbol
Adprhl1
Entrez Gene ID
234072
Gene Name
ADP-ribosylhydrolase like 1
Gene Aliases
Arh2, D330008N11Rik
Chromosome Location
Chr. 8: 13235632 - 13254263 on Build GRCm38
UniGene ID
Mm.132785
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172750.3 NP_766338.1 2 461
GenBank mRNA AJ427360.1 - 2 438
AK081786.1 - 2 295
AK084501.1 - 2 462
BC063759.1 - 2 451 30134000

View Full Product Details