Gene Transcripts

Gene Symbol
Capns1
Entrez Gene ID
12336
Gene Name
calpain, small subunit 1
Gene Aliases
Capa-4, Capa4, Capn4, D7Ertd146e
Chromosome Location
Chr. 7: 30186942 - 30198794 on Build GRCm38
UniGene ID
Mm.439782
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009795.3 NP_033925.2 2 70
GenBank mRNA AF058298.1 - 1 46
BC018352.1 - 2 64 3156021
BC090988.1 - 2 70 30246016

View Full Product Details