Gene Transcripts

Gene Symbol
Pfdn6
Entrez Gene ID
14976
Gene Name
prefoldin subunit 6
Gene Aliases
H-2Ke2, H2-Ke2, Ke-2
Chromosome Location
Chr. 17: 33938909 - 33940343 on Build GRCm38
UniGene ID
Mm.2948
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001185182.1 NP_001172111.1 3 145
NM_010385.2 NP_034515.1 2 349
GenBank mRNA AA023721.1 - 2 125
AK010679.1 - 2 339
AK076015.1 - 2 332
AK167900.1 - 2 335
AK177303.1 - 2 217
BC022974.1 - 2 315 5373335
BY238879.1 - 2 349
KJ650212.1 - 2 93
M65255.1 - 2 312

View Full Product Details